EMAIL THIS PAGE TO A FRIEND
Indian journal of pharmaceutical sciences
Validated HPLC Method for the Determination of Paclitaxel-related Substances in an Intravenous Emulsion Loaded with a Paclitaxel-Cholesterol Complex.
PMID 24591742
Abstract
A high-performance liquid chromatography method was developed for the determination of related substances in an intravenous emulsion loaded with a paclitaxel-cholesterol complex. The separation was achieved using Agilent Eclipse XDB-C18 column (150×4.6 mm, 3.5 μm), which was kept at 40°. The gradient mobile phase consisted of acetonitrile and water with a flow rate of 1.2 ml/min. The ultraviolet detection wavelength was set at 227 nm. The preparation of the sample solution began with the addition of anhydrous sodium sulphate to break the emulsion. Then, methanol and ethyl ether were added to pick up the drug and remove the accessories of the emulsion by extraction and centrifugation. Finally, paclitaxel was enriched by a nitrogen blow method and resolved with a mixture of methanol:glacial acetic acid (200:1). The method was proven to be selective, sensitive, robust, linear, repeatable, accurate and suitable for the determination of paclitaxel-related substances in the emulsion formulations, and the major degradation products in the potential pharmaceutical product were 7-epipaclitaxel and 10-deacetylpaclitaxel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8154
Baccatin III, ≥95% (HPLC)
C31H38O11