EMAIL THIS PAGE TO A FRIEND
Cell transplantation
Therapeutic effects of mouse adipose-derived stem cells and losartan in the skeletal muscle of injured mdx mice.
PMID 24593934
Abstract
Duchenne muscular dystrophy (DMD) is an X-linked genetic disorder caused by mutations in the dystrophin gene. Adipose-derived stem cells (ASCs) are an attractive source of cells for stem cell therapy. Losartan has been reported to improve ASC transplantation in injured mouse muscles. In the present study, we investigated whether the combined treatment of losartan and ASCs in the injured muscles of mdx mice improves regeneration. The combined treatment of ASCs and losartan remarkably improved muscle regeneration and induced muscle hypertrophy. In addition, ASCs and losartan treatment downregulated transforming growth factor-β and inhibited muscle fibrosis. We observed cells coexpressing green fluorescent protein (GFP) and dystrophin in the muscle samples of mice transplanted with GFP-positive ASCs. In the coculture in vitro experiment, we also observed that the GFP ASCs differentiated into dystrophin-expressing myotubes. The present study shows that the combination of transplanted ASCs and treatment with losartan ameliorated muscle fibrosis and improved muscle regeneration in injured mdx mice. Thus, we suggest that combined treatment with losartan and ASCs could help to improve muscle regeneration in the muscles of injured patients, including DMD patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500210
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500736
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4500737
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB2501687
Anti-ACPP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501605
Anti-ACPP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400002
Anti-ACPP antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA004335
Anti-ACPP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104826
Anti-ACPP antibody produced in rabbit, affinity isolated antibody
HPA002725
Anti-DMD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA023885
Anti-DMD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408565
Anti-DMD antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300474
Anti-EIF4EBP1 (Ab-36) antibody produced in rabbit, affinity isolated antibody
SAB4300475
Anti-EIF4EBP1 (Ab-45) antibody produced in rabbit, affinity isolated antibody
HPA023501
Anti-EIF4EBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106955
Anti-EIF4EBP1 antibody produced in rabbit, affinity isolated antibody
SAB4300473
Anti-FRAP1 (Ab-2448) antibody produced in rabbit, affinity isolated antibody
SAB4300583
Anti-FRAP1 (Ab-2481) antibody produced in rabbit, affinity isolated antibody
SAB2701842
Anti-mtor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701843
Anti-mtor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501038
Anti-MTOR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501039
Anti-MTOR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501040
Anti-MTOR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2501587
Anti-MYOG (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA038093
Anti-MYOG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410814
Anti-MYOG antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV32393
Anti-PAX7 (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV32742
Anti-PAX7 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1412356
ANTI-PAX7 antibody produced in mouse, clone 1G11, purified immunoglobulin, buffered aqueous solution
SAB1412376
ANTI-PAX7 antibody produced in mouse, clone 3H1, purified immunoglobulin, buffered aqueous solution
SAB1412455
ANTI-PAX7 antibody produced in mouse, clone 4F8, purified immunoglobulin, buffered aqueous solution