EMAIL THIS PAGE TO A FRIEND
British journal of clinical pharmacology
Understanding the effect of the HCV polymerase inhibitor mericitabine on early viral kinetics in the phase 2 JUMP-C and PROPEL studies.
PMID 24602156
Abstract
The aim was to evaluate early viral kinetics in patients receiving mericitabine [hepatitis C virus (HCV) nucleoside polymerase inhibitor] with peginterferon alfa-2a (40KD) and ribavirin in two clinical trials (PROPEL and JUMP-C). We examined rapid virological responses (RVRs; week 4 HCV RNA <15 IU ml(-1) ) and complete early virological responses (cEVR; week 12 HCV RNA <15 IU ml(-1) ) in HCV genotype 1/4-infected patients receiving mericitabine (500 or 1000 mg) or placebo twice daily plus peginterferon alfa-2a and ribavirin. Among IL28B rs12979860 CC genotype patients receiving 500 or 1000 mg mericitabine or placebo, respectively, RVR rates were 64.3% (95% confidence interval: 38.8-83.7%), 95.1% (83.9-98.7%) and 33.3% (20.2-49.7%), and cEVR rates were 100% (78.5-100%), 100% (91.4-100%) and 80.6% (65.0-90.3%). Among non-CC genotype patients, RVR rates were 26.5% (14.6-43.1%), 52.3% (43.0-61.3%) and 5.7% (2.2-13.8%), and cEVR rates were 76.5% (60.0-87.6%), 84.6% (76.6-90.1%) and 28.6% (19.3-40.1%), respectively. In multiple regression analysis, IL28B genotype (P < 0.0001), mericitabine dose (P < 0.0001) and bodyweight (P = 0.0009) were associated with first-phase (α) slope (change in log10 HCV RNA from baseline to week 1). Mericitabine-containing triple therapy reduces the impact of IL28B genotype on RVR and cEVR compared with peginterferon alfa-2a and ribavirin dual therapy. The IL28B genotype, mericitabine dose and bodyweight are the most important factors associated with the α slope, and there is no evidence of a pharmacokinetic drug-drug interaction between mericitabine and ribavirin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000377
Ribavirin, European Pharmacopoeia (EP) Reference Standard
C8H12N4O5