EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Reliability of computer-assisted method transfer between several column dimensions packed with 1.3-5μm core-shell particles and between various instruments.
PMID 24603352
Abstract
In this contribution, the possibility to automatically transfer RPLC methods between different column dimensions and instruments was evaluated using commercial modelling software. The method transfer reliability was tested with loratadine and its 7 related pharmacopeial impurities. In this study, state-of-the-art columns packed with superficially porous particles of 5, 2.6, 1.7 and 1.3μm particles were exclusively employed. A fast baseline separation of loratadine and related impurities (Rs,min=2.49) was achieved under the best analytical conditions (i.e. column of 50mm×2.1mm, 1.3μm, 10-90% ACN in 5min, T=40°C, pH=3, F=0.5ml/min). This optimal method was successfully tested on columns packed with other particle sizes, namely 1.7 and 2.6μm, to reduce pressure drop. The selectivities and retentions remained identical, while the peak widths were logically wider, leading to a reduction of peak capacity from 203 to 181 and 159 on the 1.3, 1.7 and 2.6μm particles, respectively. On the minimum, the resolution was equal to 1.54 on the 50mm×2.1mm, 2.6μm stationary phase. Next to this, the method was transferred to columns of different lengths, inner diameters and particle sizes (100mm×3mm, 2.6μm or 150mm×4.6mm, 5μm). These columns were used on other LC instruments possessing larger dwell volumes. The modelling software employed for developing the original method was able to calculate the new gradient conditions to be used. The accuracy of prediction was excellent, as the average retention time errors between predicted and observed chromatograms were -0.11% and 0.45% when transferring the method to 100mm×3mm and 150mm×4.6mm columns, respectively. This work proves the usefulness and validity of HPLC modelling software for transferring methods between different instruments, column dimensions and/or flow rates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9664
Loratadine, ≥98% (HPLC), powder
C22H23N2O2Cl
Y0000688
Loratadine, European Pharmacopoeia (EP) Reference Standard
C22H23N2O2Cl
1370270
Loratadine, United States Pharmacopeia (USP) Reference Standard
C22H23N2O2Cl
PHR1376
Loratadine, Pharmaceutical Secondary Standard; Certified Reference Material
C22H23N2O2Cl
Y0000603
Loratadine for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H23N2O2Cl
03366
Phosphate atomic spectroscopy standard concentrate 1.00 g PO43-, 1.00 g/L, for 1L standard solution, analytical standard
H3O4P
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
04102
Phosphoric acid, puriss., ≥99%
H3O4P
40278
Phosphoric acid, semiconductor grade MOS PURANAL (Honeywell 17938), ≥85%
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
609781
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
H316O4P
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S3139
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P
S2554
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
H2NaO4P
S6566
Sodium phosphate monobasic, Biotechnology Performance Certified, Cell Culture Tested
H2NaO4P
229903
Sodium phosphate monobasic, 99.999% trace metals basis
H2NaO4P
S0751
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
H2NaO4P
71496
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
H2NaO4P
74092
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
H2NaO4P
609773
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
H2Na16O4P