EMAIL THIS PAGE TO A FRIEND
PLoS pathogens
Chronic exposure to type-I IFN under lymphopenic conditions alters CD4 T cell homeostasis.
PMID 24603698
Abstract
HIV infection and the associated chronic immune activation alter T cell homeostasis leading to CD4 T cell depletion and CD8 T cell expansion. The mechanisms behind these outcomes are not totally defined and only partially explained by the direct cytopathic effect of the virus. In this manuscript, we addressed the impact of lymphopenia and chronic exposure to IFN-α on T cell homeostasis. In a lymphopenic murine model, this interaction led to decreased CD4 counts and CD8 T cell expansion in association with an increase in the Signal Transducer and Activator of Transcription 1 (STAT1) levels resulting in enhanced CD4 T cell responsiveness to IFN-α. Thus, in the setting of HIV infection, chronic stimulation of this pathway could be detrimental for CD4 T cell homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406025
Anti-IL7 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019590
Anti-IL7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410709
Anti-IL7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2502005
Anti-IL7R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406026
Anti-IL7R antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410710
Anti-IL7R antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1105169 Anti-STAT1 (671-685) antibody produced in rabbit, IgG fraction of antiserum
SAB4300326
Anti-STAT1 (Ab-701) antibody produced in rabbit, affinity isolated antibody
SAB4300441
Anti-STAT1 (Ab-727) antibody produced in rabbit, affinity isolated antibody
GW22758 Anti-STAT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33373
Anti-STAT1 (AB1) antibody produced in rabbit, affinity isolated antibody
GW22780 Anti-STAT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33374
Anti-STAT1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV38933
Anti-STAT1 (AB3) antibody produced in rabbit, affinity isolated antibody
AV38934
Anti-STAT1 (AB4) antibody produced in rabbit, affinity isolated antibody
SAB1406486
Anti-STAT1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000931
Anti-STAT1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36979
Anti-STAT1 antibody produced in rabbit, IgG fraction of antiserum
S5683 Anti-STAT1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous glycerol solution
HPA000982
Anti-STAT1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
RAB0316
Human IL-7 ELISA Kit, for serum, plasma, cell culture supernatant and urine
SAB4700134 Monoclonal Anti-CD27-PE antibody produced in mouse, clone LT27, purified immunoglobulin, buffered aqueous solution
SAB4700046 Monoclonal Anti-CD3-APC antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
I2532 Monoclonal Anti-Interleukin-7 antibody produced in mouse, clone 7417.111, purified immunoglobulin, lyophilized powder
WH0006772M1
Monoclonal Anti-STAT1 antibody produced in mouse, clone 1A8, purified immunoglobulin, buffered aqueous solution
RAB0317
Mouse IL-7 ELISA Kit, for serum, plasma and cell culture supernatant