EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Correlation of Sprouty1 and Jagged1 with aggressive prostate cancer cells with different sensitivities to androgen deprivation.
PMID 24604720
Abstract
Prostate cancer is a heterogeneous disease and thus, it is important to understand whether among the heterogeneous collection of cell types, androgen-deprivation insensitive cells exist prior to hormonal manipulation. We established several LNCaP subclones with distinct insensitivities to androgen deprivation from a parental LNCaP cell line. In the resulting clones, the sensitivity to androgen-deprivation negatively correlated with their PSA expression levels. In two of these clones, an androgen insensitive clone, LNCaP-cl1, and an androgen sensitive clone, LNCaP-cl5, the DNA copy number differed significantly, indicating that these clones contain genetically distinct cells. LNCaP-cl1 had higher PSA expression but lower invasiveness and tumor growth potential than LNCaP-cl5. The expression levels of two genes that are known to be regulated by miR-21, an androgen-regulated microRNA, Sprouty1 (SPRY1) and Jagged1 (JAG1) were significantly lower in LNCaP-cl1 than in LNCaP-cl5. Knocking down SPRY1 in LNCaP cells enhanced PSA expression and cell proliferation. JAG1 administration in LNCaP cells enhanced cell invasion and JAG1 knockdown in PC3 cells suppressed cell invasion and tumor formation. These results indicated that the expression differences in SPRY1 and JAG1 may contribute to the phenotypic differences between the LNCaP-cl1 and LNCaP-cl5 clones. In tissue samples, SPRY1 expression levels were significantly lower in prostate cancer patients with PSA recurrence after surgical treatment (P = 0.0076) and JAG1 expression levels were significantly higher in Gleason sum (GS) 8-9 disease than in GS 5-6 (P = 0.0121). In summary a random population of LNCaP cells comprises a heterogeneous group of cells with different androgen-deprivation sensitivities and potential for invasiveness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409018
Anti-AKR1B1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV48180
Anti-AKR1B1 antibody produced in rabbit, IgG fraction of antiserum
SAB2100091
Anti-AKR1B1 antibody produced in rabbit, affinity isolated antibody
HPA026425
Anti-AKR1B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410404
Anti-AKR1B1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1410405
Anti-AKR1B1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22855 Anti-Amphiregulin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300382
Anti-AR (Ab-650) antibody produced in rabbit, affinity isolated antibody
SAB2100139
Anti-AR antibody produced in rabbit, affinity isolated antibody
SAB1405478
Anti-AREG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA008720
Anti-AREG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410727
Anti-AREG antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1400100
Anti-FDXR antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1400101
Anti-FDXR antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2100807
Anti-FDXR antibody produced in rabbit, affinity isolated antibody
Y0001444
Bicalutamide, European Pharmacopoeia (EP) Reference Standard
C18H14F4N2O4S
EHU087521 MISSION® esiRNA, esiRNA human SPRY1 (esiRNA1)
EMU046451 MISSION® esiRNA, esiRNA targeting mouse Spry1 (esiRNA1)
SAB1402112
Monoclonal Anti-AKR1B1 antibody produced in mouse, clone 2D12, purified immunoglobulin, buffered aqueous solution
WH0000367M1
Monoclonal Anti-AR antibody produced in mouse, clone 1G3, purified immunoglobulin, buffered aqueous solution
SAB5300341 Monoclonal Anti-AR antibody produced in mouse, clone 1A9D12, ascites fluid
SAB5300342
Monoclonal Anti-AR antibody produced in mouse, clone 2H8, ascites fluid
SAB1402118
Monoclonal Anti-AREG antibody produced in mouse, clone 3E4, purified immunoglobulin, buffered aqueous solution