EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Dual platinum and pyrrolidine catalysis in the direct alkylation of allylic alcohols: selective synthesis of monoallylation products.
PMID 24604875
Abstract
A dual platinum- and pyrrolidine-catalyzed direct allylic alkylation of allylic alcohols with various active methylene compounds to produce products with high monoallylation selectivity was developed. The use of pyrrolidine and acetic acid was essential, not only for preventing undesirable side reactions, but also for obtaining high monoallylation selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
394238
Pyrrolidine, ≥99.5%, purified by redistillation
C4H9N