EMAIL THIS PAGE TO A FRIEND
Oral diseases
Characterization of mesenchymal progenitor cells in crown and root pulp from human mesiodentes.
PMID 24605962
Abstract
Mesiodentes are usually found in the central position of the upper or lower jaw as supernumerary teeth. Here, we obtained 10 mesiodentes and three permanent teeth (PT) and separated the dental pulp (DP) from these into crown and root portions. We then characterized and compared the isolated crown portion-derived cells (crown cells) with root portion-derived cells (root cells) using a range of in vitro assays. Crown cells and root cells were examined for cell surface marker expression, colony-forming unit-fibroblast (CFU-F), cell proliferation, cell cycle characteristics and markers, and osteogenic and adipogenic differentiation. The proportion of CD105-positive cells (CD105(+) cells) in the crown cells vs the root cells varied among the mesiodentes, but not among the PT. When there were more CD105(+) cells in the root cells than in the crown cells, the root cells showed higher CFU-F, proliferation capacity, and osteogenic differentiation capacity. In contrast, when the crown cells contained more CD105(+) cells than the root cells, the crown cells showed the higher CFU-F, proliferation capacity, and osteogenic differentiation capacity. In addition, the sorted CD105(+) cells showed higher CFU-F and proliferation capacity than the sorted CD105(-) cells. These results indicated that proportion of CD105(+) cells is an effective means of characterizing DP-derived cells in mesiodentes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T19607
1,2,4,5-Tetramethylbenzene, 98%
C10H14
74658
1,2,4,5-Tetramethylbenzene, Standard for quantitative NMR, TraceCERT®
C10H14
H-090
1α,25-Dihydroxyvitamin D2 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material
C28H44O3
739898
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
C28H44O3
739677
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)
C28H44O3
D1530
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
C27H44O3
17936
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
C27H44O3
A1300000
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
C6H8O6
1043003
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
C6H8O6
PHR1008
Ascorbic Acid
C6H8O6
C0225000
Calcitriol, European Pharmacopoeia (EP) Reference Standard
C27H44O3
441872
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Ca
596566
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Ca
465895
Calcium, dendritic pieces, purified by distillation, 99.5% trace metals basis
Ca
215147
Calcium, granular, 99%
Ca
327387
Calcium, pieces, <1 cm, 99%
Ca
215414
Calcium, turnings, 99% trace metals basis
Ca
I7378
Indomethacin, ≥99% (TLC)
C19H16ClNO4
I8280
Indomethacin, meets USP testing specifications
C19H16ClNO4
1341001
Indomethacin, United States Pharmacopeia (USP) Reference Standard
C19H16ClNO4
I0200000
Indomethacin, European Pharmacopoeia (EP) Reference Standard
C19H16ClNO4
PHR1247
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
C19H16ClNO4
57803
L-Ascorbic acid, certified reference material, TraceCERT®
C6H8O6
95212
L-Ascorbic acid, tested according to Ph.Eur.
C6H8O6