EMAIL THIS PAGE TO A FRIEND
Life sciences
Calcitonin gene-related peptide protects the myocardium from ischemia induced by endothelin-1: intravital microscopic observation and (31)P-MR spectroscopic studies.
PMID 24607775
Abstract
Calcitonin gene-related peptide (CGRP) is a potent vasodilator neuropeptide. We investigated the ameliorating effect of CGRP in myocardial ischemia induced by endothelin-1 (ET-1), with special emphasis on myocardial microvascular hemodynamics and levels of energy-related metabolites. The Langendorff preparations of rat isolated heart were perfused at a constant flow rate. Microvascular blood flow was also visualized in the anterior epicardium of the left ventricle by means of an intravital fluorescence microscope system. Energy-related metabolite contents in the myocardium were measured by means of (31)P-magnetic resonance spectroscopy ((31)P-MRS). Intracoronary bolus injections of CGRP caused dose-dependent decreases in coronary perfusion pressure (CPP) in the hearts exposed to ET-1 (30 pmol). The vasodilator potency of CGRP was about 10,000-fold greater than that of nitroglycerin and 1,000-fold greater than that of isobutylmethylxanthine. Vasodilation of the small-sized arterioles (10-40 μm in diameter) in response to CGRP (100 pmol) was confirmed by direct microscopic observation. After ET-1 (30 pmol) plus vehicle administration, high energy phosphates (phosphocreatine (PCr), ATP) were markedly reduced (p<0.05). CGRP administration significantly (p<0.05) attenuated the anaerobic changes in the myocardium (decrease in PCr) and macrohemodynamic alterations (increase in CPP, decrease in dP/dt etc.) induced by ET-1. We conclude that CGRP effectively confers hemodynamic and metabolic protections to isolated beating hearts against ET-1-induced myocardial ischemia.