EMAIL THIS PAGE TO A FRIEND
Nanomedicine : nanotechnology, biology, and medicine
Liposomal encapsulation of dexamethasone modulates cytotoxicity, inflammatory cytokine response, and migratory properties of primary human macrophages.
PMID 24607939
Abstract
The encapsulation of drugs into liposomes aims to enhance their efficacy and reduce their toxicity. Corticosteroid-loaded liposomes are currently being evaluated in patients suffering from rheumatoid arthritis, atherosclerosis, colitis, and cancer. Here, using several different fluorophore-labeled formulations, we comprehensively studied the impact of liposome encapsulation of the prototypic corticosteroid dexamethasone on various primary human cells in vitro. Liposomal dexamethasone targeted several primary cell types in a dose and time-dependent manner, but specifically reduced cytotoxicity against human fibroblasts and macrophages in comparison to the solute drug. Furthermore, macrophage maturation and polarization markers were altered. Interestingly, liposomal dexamethasone induced proinflammatory cytokine secretion (specifically TNF, IL1β, IL6) in unstimulated cells, but reduced this response under inflammatory conditions. Monocyte and macrophage migration was significantly inhibited by dexamethasone-loaded liposomes. The findings indicate that the encapsulation of dexamethasone into liposomes modulates their cellular mechanism of action, and provides important indications for follow-up in vivo investigations. This study investigates mechanism of action of liposomal dexamethason in the treatment of inflammatory conditions. It is concluded that liposomal dexamethasone actually induces proinflammatory cytokine secretion in unstimulated cells, but reduces the same response under inflammatory conditions. Monocyte and macrophage migration was also inhibited. The findings indicate that liposomal dexamethasone may have different mechanisms of action than its native counterpart.