EMAIL THIS PAGE TO A FRIEND
Oncogene
Surfactant protein D suppresses lung cancer progression by downregulation of epidermal growth factor signaling.
PMID 24608429
Abstract
Surfactant protein D (SP-D) is a member of the collectin family that has an important role in maintaining pulmonary homeostasis. In this study, we demonstrated that SP-D inhibited the proliferation, migration and invasion of A549 human lung adenocarcinoma cells. We found that SP-D suppressed epidermal growth factor (EGF) signaling in A549 cells, H441 human lung adenocarcinoma cells and human EGF receptor (EGFR) stable expression CHO-K1 cells. A binding study using (125)I-EGF demonstrated that SP-D downregulated the binding of EGF to EGFR. A ligand blot indicated that SP-D bound to EGFR, and a lectin blot suggested that EGFR in A549 cells had both high-mannose type and complex type N-glycans. We purified the recombinant extracellular domain of EGFR (soluble EGFR=soluble EGFR (sEGFR)), and demonstrated that SP-D directly bound to sEGFR in a Ca(2+)-dependent manner. The binding of SP-D to sEGFR was suppressed by EDTA, mannose or N-glycopeptidase F treatment. Mass spectrometric analysis indicated that N-glycans in domain III of EGFR were of a high-mannose type. These data suggest that SP-D reduces EGF binding to EGFR through the interaction between the carbohydrate recognition domain of SP-D and N-glycans of EGFR, and downregulates EGF signaling. Our finding suggests the novel type of regulation system of EGF signaling involving lectin-to-carbohydrate interaction and downregulation of ligand binding.