EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
TGF-β1 and FGF2 stimulate the epithelial-mesenchymal transition of HERS cells through a MEK-dependent mechanism.
PMID 24610459
Abstract
Hertwig's epithelial root sheath (HERS) cells participate in cementum formation through epithelial-mesenchymal transition (EMT). Previous studies have shown that transforming growth factor beta 1 (TGF-β1) and fibroblast growth factor 2 (FGF2) are involved in inducing EMT. However, their involvement in HERS cell transition remains elusive. In this study, we confirmed that HERS cells underwent EMT during the formation of acellular cementum. We found that both TGF-β1 and FGF2 stimulated the EMT of HERS cells. The TGF-β1 regulated the differentiation of HERS cells into periodontal ligament fibroblast-like cells, and FGF2 directed the differentiation of HERS cells into cementoblast-like cells. Treatment with TGF-β1 or FGF2 inhibitor could effectively suppress HERS cells differential transition. Combined stimulation with both TGF-β1 and FGF-2 did not synergistically accelerate the EMT of HERS. Moreover, TGF-β1/FGF2-mediated EMT of HERS cells was reversed by the MEK1/2 inhibitor U0126. These results suggest that TGF-β1 and FGF2 induce the EMT of HERS through a MAPK/ERK-dependent signaling pathway. They also exert their different tendency of cellular differentiation during tooth root formation. This study further expands our knowledge of tooth root morphogenesis and provides more evidence for the use of alternative cell sources in clinical treatment of periodontal diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW20078F Anti-α-2-HS-Glycoprotein antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV40173
Anti-ACTB antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405441
Anti-AHSG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001524
Anti-AHSG antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33815
Anti-AHSG antibody produced in rabbit, affinity isolated antibody
HPA001525
Anti-AHSG antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB1401138
Anti-FGF13 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2103570
Anti-FGF13 antibody produced in rabbit, affinity isolated antibody
SAB2100814
Anti-FGF2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
GW20021F Anti-Fibronectin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV41490
Anti-FN1 antibody produced in rabbit, affinity isolated antibody
HPA027066
Anti-FN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104810
Anti-FN1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1304955
ANTI-IBSP (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV36681
Anti-IBSP antibody produced in rabbit, IgG fraction of antiserum
AV33888
Anti-KLK6 antibody produced in rabbit, affinity isolated antibody
SAB4300493
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
SAB4300494
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
GW22781 Anti-MAPK3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412386
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution