EMAIL THIS PAGE TO A FRIEND
The journal of sexual medicine
An increased arginase activity is associated with corpus cavernosum impairment induced by hypercholesterolemia.
PMID 24612630
Abstract
Hypercholesterolemia is a prevalent risk factor for the development of erectile dysfunction (ED), mostly due to an increase in oxidative stress and impaired nitric oxide (NO) bioavailability within the penis. Arginase is an enzyme that shares the common substrate L-arginine with NO synthase. Augmented arginase activity reduces NO production and is associated with ED development. However, the contribution of arginase hyperactivity in hypercholesterolemia-induced ED is unknown. In the present study, we investigated the activity and role of arginase in the corpus cavernosum of hypercholesterolemic mice. Apolipoprotein E (ApoE) gene-deleted mice fed with a Western-type diet for 11 weeks were treated with the selective arginase inhibitor, N-ω-Hydroxy-L-norarginine (NOHA), or vehicle (saline 0.9%) during the last 9 weeks. Arginase activity and expression were measured in penis protein extraction. Reactive oxygen species (ROS) content within the corpus cavernosum was measured by dihydroethidium staining. Functional in vitro studies were performed using cavernosal strips mounted in an isometric organ bath to evaluate NO production. Arginase activity and its role in modulating NO and ROS production within the corpus cavernosum of hypercholesterolemic mice is the main outcome measure. Total arginase activity and arginase type II protein expression were increased in hypercholesterolemic mice compared with wild-type mice. The long-term treatment with NOHA normalized this alteration. Moreover, pharmacological arginase inhibition by NOHA attenuated the augmented ROS production within the corpus cavernosum of ApoE(-/-) mice, which increased the NO-dependent response in cavernosal strips. These evidences indicate that arginase hyperactivity is associated with ED induced by hypercholesterolemia, suggesting that this enzyme is a potential target for treating ED.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV45673
Anti-ARG1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1405480
Anti-ARG1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003595
Anti-ARG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024006
Anti-ARG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100144
Anti-ARG1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4200510
Anti-ARG1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1405481
Anti-ARG2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000663
Anti-ARG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV54567
Anti-ARG2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1410736
Anti-ARG2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
D7008
Dihydroethidium, ≥95%
C21H21N3
37291
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
C21H21N3
SAB4200508
Monoclonal Anti-ARG1 antibody produced in mouse, ~1.0 mg/mL, clone ARG1-3, purified immunoglobulin
SAB4100251
Monoclonal Anti-ARG1 antibody produced in mouse, clone ARG1-A219, culture supernatant
SAB4200509
Monoclonal Anti-ARG1 antibody produced in mouse, ~1.0 mg/mL, clone ARG1-6, purified immunoglobulin
AMAB90545
Monoclonal Anti-ARG1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0186, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402693
Monoclonal Anti-ARG2 antibody produced in mouse, clone 3D10, purified immunoglobulin, buffered aqueous solution