EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Altered lipid composition in cortical lipid rafts occurs at early stages of sporadic Alzheimer's disease and facilitates APP/BACE1 interactions.
PMID 24613671
Abstract
The presence of lipid alterations in lipid rafts from the frontal cortex in late stages of Alzheimer's disease (AD) has been recently demonstrated. Here, we have isolated and analyzed the lipid composition of lipid rafts from different brain areas from control and AD subjects at initial neuropathologic stages. We have observed that frontal cortex lipid rafts are profoundly altered in AD brains from the earliest stages of AD, namely AD I/II. These changes in the lipid matrix of lipid rafts affected both lipid classes and fatty acids and were also detected in the entorhinal cortex, but not in the cerebellum from the same subjects. Paralleling these changes, lipid rafts from AD frontal and entorhinal cortices displayed higher anisotropy for environment-sensitive probes, indicating that lipid changes in AD lipid rafts increased membrane order and viscosity in these domains. The pathophysiological consequences of these alterations in the development and progression of AD were strengthened by the significant, and specific, accumulation of β-secretase within the lipid rafts of AD subjects even at the earliest stages. Our results provide a mechanistic connection between lipid alterations in these microdomains and amyloidogenic processing of amyloid precursor protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D208000
1,6-Diphenyl-1,3,5-hexatriene, 98%
C18H16
SAB4300464
Anti-APP (Ab-668) antibody produced in rabbit, affinity isolated antibody
SAB2100134
Anti-APP (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100135
Anti-APP (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100136
Anti-APP (ab3) antibody produced in rabbit, IgG fraction of antiserum
HPA001462
Anti-APP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200535
Anti-APP antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB3500274
Anti-APP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB5200113
Anti-APP antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
SAB2107345
Anti-APP antibody produced in rabbit, affinity isolated antibody
SAB2106270
Anti-APP antibody produced in rabbit, affinity isolated antibody
SAB2900061
Anti-CAV1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900062
Anti-CAV1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV09019
Anti-CAV1 antibody produced in rabbit, purified antibody, Protein A purified
SAB2900063
Anti-CAV1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100344
Anti-CAV1 antibody produced in rabbit, affinity isolated antibody
C4490
Anti-Caveolin-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
C3237
Anti-Caveolin-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2701919 Anti-flot1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001393
Anti-FLOT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410072
Anti-FLOT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500404
Anti-Flotillin 1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F1180
Anti-Flotillin 1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
P5110 Anti-Presenilin-1 (303-316) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
P7854
Anti-Presenilin-1 (S182) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA030760
Anti-PSEN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502423
Anti-PSEN1 antibody produced in rabbit, affinity isolated antibody
SAB1403056
Monoclonal Anti-FLOT1 antibody produced in mouse, clone 4A1, ascites fluid