EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
The Ca²⁺-calmodulin-Ca²⁺/calmodulin-dependent protein kinase II signaling pathway is involved in oxidative stress-induced mitochondrial permeability transition and apoptosis in isolated rat hepatocytes.
PMID 24614978
Abstract
Oxidative stress (OS) is a common event in most hepatopathies, leading to mitochondrial permeability transition pore (MPTP) formation and further exacerbation of both OS from mitochondrial origin and cell death. Intracellular Ca²⁺ increase plays a permissive role in these events, but the underlying mechanisms are poorly known. We examined in primary cultured rat hepatocytes whether the Ca²⁺/calmodulin (CaM)-dependent protein kinase II (CaMKII) signaling pathway is involved in this process, by using tert-butyl hydroperoxide (tBOOH) as a pro-oxidant, model compound. tBOOH (500 μM, 15 min) induced MPTP formation, as assessed by measuring mitochondrial membrane depolarization as a surrogate marker, and increased lipid peroxidation in a cyclosporin A (CsA)-sensitive manner, revealing the involvement of MPTPs in tBOOH-induced radical oxygen species (ROS) formation. Intracellular Ca²⁺ sequestration with BAPTA/AM, CaM blockage with W7 or trifluoperazine, and CaMKII inhibition with KN-62 all fully prevented tBOOH-induced MPTP opening and reduced tBOOH-induced lipid peroxidation to a similar extent to CsA, suggesting that Ca²⁺/CaM/CaMKII signaling pathway fully mediates MPTP-mediated mitochondrial ROS generation. tBOOH-induced apoptosis, as shown by flow cytometry of annexin V/propidium iodide, mitochondrial release of cytochrome c, activation of caspase-3 and increase in the Bax-to-Bcl-xL ratio, and the Ca²⁺/CaM/CaMKII signaling antagonists fully prevented these effects. Intramitochondrial CaM and CaMKII were partially involved in tBOOH-induced MPTP formation, since W7 and KN-62 both attenuated the tBOOH-induced, MPTP-mediated swelling of isolated mitochondria. We concluded that Ca²⁺/CaM/CaMKII signaling pathway is a key mediator of OS-induced MPTP formation and the subsequent exacerbation of OS from mitochondrial origin and apoptotic cell death.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14510
1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid, ≥96.0% (HPLC)
C22H24N2O10
B3428 Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500343
Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA027878
Anti-BAX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502549
Anti-BAX antibody produced in rabbit, affinity isolated antibody
B8429
Anti-Bax antibody, Mouse monoclonal, clone 6A7, purified from hybridoma cell culture
B8554
Anti-Bax antibody, Mouse monoclonal, clone 2D2, purified from hybridoma cell culture
SAB2101602
Anti-NKX3-1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101603
Anti-NKX3-1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1412056
ANTI-NKX3-1 antibody produced in mouse, clone 3C1, purified immunoglobulin, buffered aqueous solution
32425
Cyclosporin A, VETRANAL, analytical standard
C62H111N11O12
PHR1092
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
C62H111N11O12
I2142
KN-62, ≥95%, powder
C38H35N5O6S2
458139
Luperox® TBH70X, tert-Butyl hydroperoxide solution, 70 wt. % in H2O
C4H10O2
B9054 Monoclonal Anti-Bax antibody produced in mouse, 2 mg/mL, clone 5B7, purified immunoglobulin, buffered aqueous solution
WH0000581M1
Monoclonal Anti-BAX antibody produced in mouse, clone 1F5-1B7, purified immunoglobulin, buffered aqueous solution
WH0004824M2
Monoclonal Anti-NKX3-1 antibody produced in mouse, clone 1C7, purified immunoglobulin, buffered aqueous solution
SAB1404130
Monoclonal Anti-NKX3-1 antibody produced in mouse, clone 3H6, purified immunoglobulin, buffered aqueous solution
SAB5300411
Monoclonal Anti-NKX3-1 antibody produced in mouse, clone 4H4, ascites fluid
1642802
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
C44H69NO12 · H2O
19997
tert-Butyl hydroperoxide solution, packed in FEP bottles, ~5.5 M in decane (over molecular sieve 4Å)
C4H10O2
416665
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
C4H10O2
19999
tert-Butyl hydroperoxide solution, packed in FEP bottles, ~5.5 M in nonane (over molecular sieve 4 Å)
C4H10O2
418064
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
C4H10O2