EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Synthesis and antitumor activities of some new N1-(flavon-6-yl)amidrazone derivatives.
PMID 24615985
Abstract
A new series of N1-(flavon-6-yl)amidrazones were synthesized by reacting the hydrazonoyl chloride derived from 6-aminoflavone with the appropriate sec-cyclic amines. The antitumor activities of these compounds were evaluated on breast cancer (MCF-7) and leukemic (K562) cell lines. Among the compounds tested, the N-morpholine derivative was the most active against the MCF-7 and K562 cell lines, with IC50 values of 5.18 and 2.89 μM, respectively. Our docking studies showed that the N-morpholino derivative fits and blocks the oncogenic tyrosine kinases bcr/abl and epidermal growth factor receptor (EGFR) in a similar fashion to that of the potent anticancer agent imatinib.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14549
1-Methylpiperazine, analytical standard
C5H12N2
130001
1-Methylpiperazine, 99%
C5H12N2
10872
3-Chloro-2,4-pentanedione, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
C5H7ClO2
227153
3-Chloro-2,4-pentanedione, 97%
C5H7ClO2
576263
6-Aminoflavone, 97%
C15H11NO2
Y0000886
Cyclizine impurity A, European Pharmacopoeia (EP) Reference Standard
C5H12N2
394467
Morpholine, purified by redistillation, ≥99.5%
C4H9NO
252360
Morpholine, ACS reagent, ≥99.0%
C4H9NO
134236
Morpholine, ReagentPlus®, ≥99%
C4H9NO
90179
Morpholine, analytical standard
C4H9NO
493813
Morpholine, polymer-bound, 200-400 mesh, extent of labeling: 2.75-3.25 mmol/g loading, 1 % cross-linked
C29H33NO
80621
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
C4H10N2
94209
Piperazine, analytical standard
C4H10N2
80620
Piperazine, anhydrous, ≥99.0% (T)
C4H10N2
P45907
Piperazine, ReagentPlus®, 99%
C4H10N2
W425001
Piperazine, 99%
C4H10N2
411027
Piperidine, ≥99.5%, purified by redistillation
C5H11N
104094
Piperidine, ReagentPlus®, 99%
C5H11N
76046
Piperidine, analytical standard
C5H11N
80642
Piperidine, purum, ≥96.0% (GC/T)
C5H11N
W290807
Piperidine, ≥99%, FCC
C5H11N
80645
Piperidine solution, for peptide synthesis, 20% in DMF
C5H11N
547522
StratoSpheres PL-MPH resin, 50-100 mesh, extent of labeling: 3.0-4.0 mmol/g loading, 1 % cross-linked
C29H33NO
196274
Thiomorpholine, 98%
C4H9NS