EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Human RECQL1 participates in telomere maintenance.
PMID 24623817
Abstract
A variety of human tumors employ alternative and recombination-mediated lengthening for telomere maintenance (ALT). Human RecQ helicases, such as BLM and WRN, can efficiently unwind alternate/secondary structures during telomere replication and/or recombination. Here, we report a novel role for RECQL1, the most abundant human RecQ helicase but functionally least studied, in telomere maintenance. RECQL1 associates with telomeres in ALT cells and actively resolves telomeric D-loops and Holliday junction substrates. RECQL1 physically and functionally interacts with telomere repeat-binding factor 2 that in turn regulates its helicase activity on telomeric substrates. The telomeric single-stranded binding protein, protection of telomeres 1 efficiently stimulates RECQL1 on telomeric substrates containing thymine glycol, a replicative blocking lesion. Loss of RECQL1 results in dysfunctional telomeres, telomere loss and telomere shortening, elevation of telomere sister-chromatid exchanges and increased aphidicolin-induced telomere fragility, indicating a role for RECQL1 in telomere maintenance. Further, our results indicate that RECQL1 may participate in the same pathway as WRN, probably in telomere replication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406382
Anti-RECQL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401334
Anti-RECQL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA030960
Anti-RECQL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV35751
Anti-TBPL1 antibody produced in rabbit, IgG fraction of antiserum
SAB2700856
Anti-TBPL1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406522
Anti-TERF2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001907
Anti-TERF2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV30228
Anti-TERF2 antibody produced in rabbit, IgG fraction of antiserum
HPA002735 Anti-TERF2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Y0000119
Hydroxycarbamide, European Pharmacopoeia (EP) Reference Standard
CH4N2O2
EHU054161 MISSION® esiRNA, esiRNA human TBPL1 (esiRNA1)
EHU042991 MISSION® esiRNA, esiRNA human TERF2 (esiRNA1)
EHU087561 MISSION® esiRNA, esiRNA human RECQL (esiRNA1)
EMU009421 MISSION® esiRNA, esiRNA targeting mouse Wrn (esiRNA1)
EHU048411 MISSION® esiRNA, esiRNA human WRN (esiRNA1)
EMU001061 MISSION® esiRNA, esiRNA targeting mouse Terf2 (esiRNA1)
EMU089031 MISSION® esiRNA, esiRNA targeting mouse Recql (esiRNA1)
EMU040131 MISSION® esiRNA, esiRNA targeting mouse Tbpl1 (esiRNA1)
PLA0071
Rabbit anti-Phospho RPA32 (S4/S8) Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0144
Rabbit anti-TRF2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.