EMAIL THIS PAGE TO A FRIEND
Immunology
Evaluation of peroxisome proliferator-activated receptor agonists on interleukin-5-induced eosinophil differentiation.
PMID 24628018
Abstract
Peroxisome proliferator-activated receptor (PPAR) agonists have been suggested as novel therapeutics for the treatment of inflammatory lung disease, such as allergic asthma. Treatment with PPAR agonists has been shown to inhibit airway eosinophilia in murine models of allergic asthma, which can occur through several mechanisms including attenuated generation of chemoattractants (e.g. eotaxin) and decreased eosinophil migrational responses. In addition, studies report that PPAR agonists can inhibit the differentiation of several cell types. To date, no studies have examined the effects of PPAR agonists on interleukin-5 (IL-5) -induced eosinophil differentiation from haemopoietic progenitor cells. Non-adherent mononuclear cells or CD34(+) cells isolated from the peripheral blood of allergic subjects were grown for 2xa0weeks in Methocult(®) cultures with IL-5 (10xa0ng/ml) and IL-3 (25xa0ng/ml) in the presence of 1-1000xa0nm PPARα agonist (GW9578), PPARβ/δ agonist (GW501516), PPARγ agonist (rosiglitazone) or diluent. The number of eosinophil/basophil colony-forming units (Eo/B CFU) was quantified by light microscopy. The signalling mechanism involved was assessed by phosphoflow. Blood-extracted CD34(+) cells cultured with IL-5 or IL-5xa0+xa0IL-3 formed Eo/B CFU, which were significantly inhibited by rosiglitazone (100xa0nm, Pxa0