EMAIL THIS PAGE TO A FRIEND
Cancer research
Membrane-associated interleukin 1 alpha as a mediator of tumor cell killing by human blood monocytes fixed with paraformaldehyde.
PMID 2463072
Abstract
Human blood monocytes isolated by centrifugal elutriation from healthy donors were tested for ability to produce membrane-associated antitumor monokine(s) in response to activation stimuli such as various types of interferon (IFN) and/or synthetic desmethyl muramyl dipeptide (norMDP). IFNs (alpha, beta, and gamma) and norMDP rendered blood monocytes cytotoxic to allogeneic A375 melanoma cells, as assayed by measuring release of [125I]iododeoxyuridine in 72 h. When monocytes were treated with any type of IFN for 16 h, and then fixed with paraformaldehyde, they did not show cytotoxicity to A375 cells, but when they were fixed after treatment with norMDP or lipopolysaccharide they showed significant cytotoxicity to A375 melanoma cells. This membrane-associated antitumor monokine induced by the synergistic actions of suboptimal concentrations of IFN-gamma and norMDP, was cytotoxic to HT-29 colon cancer cells as well as A375 melanoma cells, but not to actinomycin D-treated L-929 cells. The fixed monocyte-mediated cytotoxicity against A375 melanoma cells was completely inhibited by a specific anti-interleukin 1 alpha antiserum, but not by a specific anti-interleukin 1 beta antiserum or monoclonal anti-TNF antibody. These results suggest that membrane-associated interleukin 1 alpha is involved through cell-to-cell contact in the host defense mechanism against cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88113005
A375 Cell Line human