EMAIL THIS PAGE TO A FRIEND
Neurobiology of disease
Improved neuroprotection using miglustat, curcumin and ibuprofen as a triple combination therapy in Niemann-Pick disease type C1 mice.
PMID 24631719
Abstract
Niemann-Pick disease type C (NPC) is a neurodegenerative lysosomal storage disorder characterised by the storage of multiple lipids, reduced lysosomal calcium levels, impaired late endosome:lysosome fusion and neuroinflammation. NPC is caused by mutations in either of the two genes, NPC1 or NPC2, which are believed to function in a common cellular pathway, the function of which remains unclear. The complexity of the pathogenic cascade in NPC disease provides a number of potential clinical intervention points. To date, drugs that target pivotal stages in the pathogenic cascade have been tested as monotherapies or in combination with a second agent, showing additive or synergistic benefit. In this study, we have investigated whether we can achieve greater therapeutic benefit in the Npc1(-/-) mouse by combining three therapies that each targets unique aspects of the pathogenic cascade. We have treated Npc1(-/-) mice with miglustat that targets sphingolipid synthesis and storage, curcumin that compensates for the lysosomal calcium defect by elevating cytosolic calcium, and the non-steroidal anti-inflammatory drug ibuprofen to reduce central nervous system inflammation. We have found that triple combination therapy has a greater neuroprotective benefit compared with single and dual therapies, increasing the time period that Npc1(-/-) mice maintained body weight and motor function and maximally delaying the onset of Purkinje cell loss. In addition, ibuprofen selectively reduced microglial activation, while curcumin had no anti-inflammatory effects, indicating differential mechanisms of action for these two therapies. When taken together, these results demonstrate that targeting multiple unique steps in the pathogenic cascade maximises the clinical benefit in a mouse model of NPC1 disease.