EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
EZH2 mutations in follicular lymphoma from different ethnic groups and associated gene expression alterations.
PMID 24634383
Abstract
Gain-of-function mutations of enhancer of Zeste homolog 2 (EZH2) occur frequently in diffuse large B-cell lymphomas and in follicular lymphomas. However, the frequency of EZH2 mutation in Chinese follicular lymphomas and the potential targets affected by this mutation are unknown. We determined EZH2 codon 641 mutations in Chinese follicular lymphomas (n = 124) and compared them with Western follicular lymphomas (n = 70) using a sensitive pyrosequencing assay. Gene expression profiling (GEP) was performed to determine differential gene expression between the mutated versus unmutated subgroups, and selected genes were validated using immunohistochemistry. Our results showed similar frequencies of EZH2 codon 641 mutations in Chinese and Western follicular lymphoma cohorts (16.9% vs. 18.6%, χ(2) test, P = 0.773), including all five reported mutation variants. We observed significant association of EZH2 mutation with low morphologic grade follicular lymphomas (grade 1-2, 23.6% vs. grade 3, 7.7%, χ(2) test, P = 0.02). EZH2 mutations also showed significant association with BCL2 rearrangement in the Chinese cohort (26.8% vs. 8.8%, χ(2) test, P = 0.008) and combined cohorts (26.3% vs. 9.1%, χ(2) test, P = 0.002). GEP analysis identified several genes, including TCF4, FOXP1, TCL1A, BIK, and RASSF6P, with significantly lower mRNA expression (P < 0.01) in mutated cases, and the potential target TCL1A showed consistent results at the protein level. Similar prevalence of EZH2 mutation in two ethnic groups suggests shared pathogenetic mechanisms. The much lower frequency of EZH2 mutation in cases without BCL2 translocation suggests a different pattern of evolution of this subtype of follicular lymphoma. GEP studies showed a set of differentially expressed genes and suggested that EZH2 mutation may help to lock the tumor cells at the germinal center stage of differentiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22194F Anti-TCL1A antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501025
Anti-TCL1A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406645
Anti-TCL1A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411982
ANTI-TCL1A antibody produced in mouse, clone 2F1, purified immunoglobulin, buffered aqueous solution
SAB1412201
ANTI-TCL1A antibody produced in mouse, clone 3H6, purified immunoglobulin, buffered aqueous solution
SAB1412206
ANTI-TCL1A antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
SAB1412230
ANTI-TCL1A antibody produced in mouse, clone 1E8, purified immunoglobulin, buffered aqueous solution
SAB1412383
ANTI-TCL1A antibody produced in mouse, clone 1G10, purified immunoglobulin, buffered aqueous solution
SAB1412384
ANTI-TCL1A antibody produced in mouse, clone 3A4, purified immunoglobulin, buffered aqueous solution
HPA016604
Anti-TCL1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104341
Anti-TCL1A, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0008115M6
Monoclonal Anti-TCL1A antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution