EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
Systematic review and metaanalysis on nonclassic cardiovascular biomarkers after hypertensive pregnancy disorders.
PMID 24637129
Abstract
The aim of this study was to investigate which nonclassic cardiovascular biomarkers are associated with persistent endothelial dysfunction after pregnancy in women with a history of hypertensive pregnancy disorders compared with women with uncomplicated pregnancies. This was a systematic review and metaanalysis of observational studies. A search was performed in PubMed, Embase, Cochrane, and Cinahl including articles from inception to Feb. 27, 2013. Included were cohort studies and case-control studies. Cases were women with a history of hypertension in pregnancy, control subjects were women with a history of uncomplicated pregnancies. Of the 3136 found, 21 studies on 16 nonclassic cardiovascular biomarkers are described in this review; 12 studies on 5 biomarkers were included in the metaanalysis. Women with a history of hypertensive pregnancy disorders had a higher homocysteine level compared with women with a history of uncomplicated pregnancies (5 studies; pooled mean difference, 0.77 ng/mL; 95% confidence interval, 0.27-1.26; P < .01). For the other nonclassic cardiovascular biomarkers including markers in areas of inflammation, thrombosis, and angiogenesis, we found no significant differences. This review and metaanalysis showed that women with a history of hypertensive pregnancy disorders have higher homocysteine levels compared with women with a history of uncomplicated pregnancies. These data suggest persistent endothelial alteration after pregnancies complicated by hypertensive disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69453
L-Homocysteine, ≥98.0% (NT)
C4H9NO2S