EMAIL THIS PAGE TO A FRIEND
Stem cells and development
Involvement of hepatic stimulator substance in the regulation of hepatoblast maturation into hepatocytes in vitro.
PMID 24640968
Abstract
Hepatic stimulator substance (HSS), also known as augmenter of liver regeneration (ALR), acts as a hepatotrophic growth factor to promote liver regeneration after liver damage or partial hepatectomy. However, the expression and function of HSS during liver development in mammals remain largely unknown. In this work, the hepatoblasts were isolated from mice at embryonic day 13.5 (E13.5), and HSS expression and its role during hepatoblast maturation were investigated. The results showed that HSS expression was enhanced in the hepatoblasts compared with mouse primary hepatocytes. HSS expression (23 kDa) was significantly decreased if the hepatoblast maturation was induced by a combination of oncostatin M (OSM), dexamethasone (DEX), and hepatocyte growth factor (HGF). We also found that knockdown of HSS expression (mainly 23-kDa isoform) by siRNA promoted hepatoblast maturation and also activated the signal transducer and activator of transcription 3 (STAT3) phosphorylation levels. However, if STAT3 activity was blocked by a small-molecule inhibitor Stattic, then hepatocyte maturation could be abolished, suggesting that STAT3 was most likely a potential molecule responsible for HSS signaling. In summary, our results demonstrated for the first time that HSS might be an active factor participating in the regulation of liver development and hepatocyte maturation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406973
Anti-AKR1A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA017919
Anti-AKR1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA019649
Anti-AKR1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA027734
Anti-AKR1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031443
Anti-EXOC4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104227
Anti-EXOC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1304975
ANTI-GFER (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA041227
Anti-GFER antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501670
Anti-Keratin 19 antibody produced in rabbit, affinity isolated antibody
SAB4300551
Anti-KRT18 (Ab-33) antibody produced in rabbit, affinity isolated antibody
AV40205
Anti-KRT18 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV40206
Anti-KRT18 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22753 Anti-KRT18 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2701928
Anti-krt18 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101302
Anti-KRT19 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101303
Anti-KRT19 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1400148
Anti-KRT19 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA002465 Anti-KRT19 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0010327M1
Monoclonal Anti-AKR1A1 antibody produced in mouse, clone 1A11-2A4, purified immunoglobulin, buffered aqueous solution
SAB4700725 Monoclonal Anti-CD117-APC antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
SAB3300015
Monoclonal Anti-CK18 antibody produced in mouse, clone 10, purified immunoglobulin, buffered aqueous glycerol solution
SAB3300017 Monoclonal Anti-CK18 antibody produced in mouse, clone 1H10, purified immunoglobulin, buffered aqueous glycerol solution
SAB3300019
Monoclonal Anti-CK19 antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700775
Monoclonal Anti-Cytokeratin 19 antibody produced in mouse, clone A53-B/A2, purified immunoglobulin, buffered aqueous solution
WH0060412M6
Monoclonal Anti-EXOC4 antibody produced in mouse, clone 4F1, purified immunoglobulin, buffered aqueous solution
WH0003875M1
Monoclonal Anti-KRT18 antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution
SAB1404012
Monoclonal Anti-KRT18 antibody produced in mouse, clone 1G8, purified immunoglobulin, buffered aqueous solution
SAB4700411
Monoclonal Anti-KRT18 antibody produced in mouse, clone C-04, purified immunoglobulin, buffered aqueous solution
SAB4700670 Monoclonal Anti-KRT18 antibody produced in mouse, clone DC-10, purified immunoglobulin, buffered aqueous solution