EMAIL THIS PAGE TO A FRIEND
Nephrology news & issues
Toxic effects of IV iron preparations in CKD patients.
PMID 24649744
Abstract
Loss of blood associated with hemodialysis procedures and laboratory testing, together with impaired iron absorption due to elevation of hepcidin, invariably cause iron deficiency in end-stage renal disease patients. For this reason, nearly all ESRD patients require intravenous iron to replete iron stores. Unfortunately, intravenously administered iron is often used routinely with inadequate attention to the body iron stores or severity of systemic inflammation. This has led to an epidemic of iron overload in the ESRD population. Only a minute amount (3-4 mg) of the total body iron (3-4 g in an adult man) resides in the plasma bound to transferrin, which serves as a safe vehicle for iron transport in the circulation. IV iron products are generally administered as bolus injections of 100 to 1000 mg, which far exceeds the available pool of free transferrin and represents a huge quantity compared to the intestinal iron absorption of 1 to 2 mg/day in the course of 3 to 4 meals. Administration of these products results in an increased plasma level of catalytically active non-transferrin bound iron and the rise in the biomarkers of oxidative stress and inflammation. IV iron bypasses the biological safeguards for the transport and handling of iron and helps to intensify chronic kidney disease-associated oxidative stress and inflammation. As briefly described in this review, indiscriminate use of IV iron can accelerate cardiovascular disease, promote microbial infections, aggravate viral hepatitis, and worsen diabetes and diabetic complications in such patients. For these reasons IV iron should be used judiciously in this vulnerable population.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3518
Carbonyl iron, ≥97% Fe basis
Fe
44890
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Fe
12310
Iron, ≥99%, reduced, powder (fine)
Fe
267945
Iron, chips, 99.98% trace metals basis
Fe
338141
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Fe
356808
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Fe
413054
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Fe
513423
Iron, activated, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 99.5% trace metals basis
Fe
IRMM524A
Iron, IRMM® certified Reference Material, 0.1 mm foil
Fe
IRMM524B
Iron, IRMM® certified Reference Material, 0.5 mm wire
Fe
255637
Iron, powder, ≥99.99% trace metals basis
Fe
209309
Iron, powder, −325 mesh, 97%
Fe
267953
Iron, powder, <10 μm, ≥99.9% trace metals basis
Fe
266213
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Fe
266256
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Fe
266248
Iron, wire, diam. 0.5 mm, ≥99.99%
Fe
356824
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Fe
356832
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Fe
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF34071566
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Fe
GF96915134
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Fe
GF35663935
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Fe
GF27511781
Iron, wire reel, 5m, diameter 0.25mm, annealed, 99.99+%
Fe
GF61040023
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Fe