EMAIL THIS PAGE TO A FRIEND
Thrombosis and haemostasis
Interruption of classic CD40L-CD40 signalling but not of the novel CD40L-Mac-1 interaction limits arterial neointima formation in mice.
PMID 24652469
Abstract
The co-stimulatory immune molecule CD40L figures prominently in a variety of inflammatory conditions including arterial disease. Recently, we made the surprising finding that CD40L mediates atherogenesis independently of its classic receptor CD40 via a novel interaction with the leukocyte integrin Mac-1. Here, we hypothesised that selective blockade of the CD40L-Mac-1 interaction may also retard restenosis. We induced neointima formation in C57/BL6 mice by ligation of the left carotid artery. Mice were randomised to daily intraperitoneal injections of either cM7, a small peptide selectively inhibiting the CD40L-Mac-1 interaction, scM7, a scrambled control peptide, or saline for 28 days. Interestingly, cM7-treated mice developed neointima of similar size compared with mice receiving the control peptide or saline as assessed by computer-assisted analysis of histological cross sections. These data demonstrate that the CD40L-Mac-1 interaction is not required for the development of restenosis. In contrast, CD40-deficient mice subjected to carotid ligation in parallel, developed significantly reduced neointimal lesions compared with respective wild-type controls (2872 ± 843 µm² vs 35469 ± 11870 µm²). Flow cytometry in CD40-deficient mice revealed reduced formation of platelet-granulocyte and platelet-inflammatory monocyte- aggregates. In vitro, supernatants of CD40-deficient platelet-leukocyte aggregates attenuated proliferation and increased apoptosis of smooth muscle cells. Unlike in the setting of atherosclerosis, CD40L mediates neointima formation via its classic receptor CD40 rather than via its recently described novel interaction with Mac-1. Therefore, selective targeting of CD40L-Mac-1 binding does not appear to be a favorable strategy to fight restenosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1305743
ANTI-CD106 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300383
Anti-ICAM1 (Ab-512) antibody produced in rabbit, affinity isolated antibody
SAB2502120
Anti-ICAM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405977
Anti-ICAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002126
Anti-ICAM1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004877
Anti-ICAM1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406579
Anti-VCAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
RAB0365
Human MMP-2 ELISA Kit, for serum, plasma, cell culture supernatant
RAB0372
Human MMP-9 ELISA Kit, for serum, plasma, cell culture supernatant and urine
SAB4700521 Monoclonal Anti-CD106 antibody produced in mouse, clone STA, purified immunoglobulin, buffered aqueous solution
SAB1305520
MONOCLONAL ANTI-CD106 antibody produced in mouse, clone 582CT25.6.4, crude ascites, buffered aqueous solution
SAB4700326 Monoclonal Anti-CD54 antibody produced in mouse, clone MEM-112, purified immunoglobulin, buffered aqueous solution
SAB4700327
Monoclonal Anti-CD54 antibody produced in mouse, clone MEM-111, purified immunoglobulin, buffered aqueous solution
SAB4700571 Monoclonal Anti-CD54 antibody produced in rat, clone YN1/1.7.4, purified immunoglobulin, buffered aqueous solution
WH0003383M1
Monoclonal Anti-ICAM1 antibody produced in mouse, clone 3H8-2G6, purified immunoglobulin, buffered aqueous solution
V9263 Monoclonal Anti-Vascular Cell Adhesion Molecule 1 antibody produced in mouse, ~1 mg/mL, clone 1.4C3, purified immunoglobulin, buffered aqueous solution
SAB1402959
Monoclonal Anti-VCAM1 antibody produced in mouse, clone 1H6, purified immunoglobulin, buffered aqueous solution
SAB4100447
Monoclonal Anti-VCAM1 antibody produced in mouse, clone VCAM1-B382, culture supernatant
RAB0366
Mouse MMP-2 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0373
Mouse pro-MMP-9 ELISA Kit, for serum, plasma and cell culture supernatant