EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Pharmacokinetics of honokiol after intravenous guttae in beagle dogs assessed using ultra-performance liquid chromatography-tandem mass spectrometry.
PMID 24652775
Abstract
A simple, rapid and sensitive ultra-performance liquid chromatography-tandem mass spectrometry method was developed and validated for the determination of honokiol in beagle dog plasma after intravenous guttae. With addition of the internal standard magnolol, plasma samples were precipitated with methanol and separated on a Shim-pack XR-ODS II (2.0 × 100 mm, 2.2 µm) with isocratic elution of methanol and water (80:20) solution at a flow rate of 0.2 mL/min. A good separation of honokiol was achieved within 3.5 min. Quantification was performed on a Waters Quattro Premier XE triple quadrupole mass spectrometer with electrospray ionization inlet in the negative multiple reaction monitoring mode. Good linearity was obtained over the concentration range of 5.12-15580 ng/mL (r(2) > 0.998). Intra- and inter-day precisions were <13.10%, and accuracy ranged from 89.21 to 99.92%. The lower limit of quantification for honokiol was 5.12 ng/mL, and honokiol was stable under various conditions (three freeze-thaw cycles, short-term temperature, post-preparative and long-term temperature conditions.). This validated method was successfully applied to the pharmacokinetic study of honokiol in dogs by intravenous guttae.