EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Targeting of BRAF resistant melanoma via extracellular matrix metalloproteinase inducer receptor.
PMID 24655664
Abstract
The BRAF inhibitor vemurafenib (PLX) has shown promise in treating metastatic melanoma, but most patients develop resistance to treatment after 6xa0mo. We identified a transmembrane protein, extracellular matrix metalloproteinase inducer (EMMPRIN) as a cell surface receptor highly expressed by PLX-resistant melanoma. Using an S100A9 ligand, we created an EMMPRIN targeted probe and liposome that binds to melanoma cells inxa0vivo, thus designing a novel drug delivery vehicle. PLX-resistant cells were established through continuous treatment with PLX-4032 over the course of 1xa0y. Both PLX-resistant and sensitive melanoma cell lines were evaluated for the expression of unique cell surface proteins, which identified EMMPRIN as an overexpressed protein in PLX0-resistant cells and S100A9 is a ligand for EMMPRIN. To design a probe for EMMPRIN, S100A9 ligand was conjugated to a CF-750 near-infrared (NIR) dye. EMMPRIN targeted liposomes were created to encapsulate CF-750 NIR dye. Liposomes were characterized by scanning electron microscopy, flow cytometry, and inxa0vivo analysis. A2058PLX and A2058 cells were subcutaneously injected into athymic mice. S100A9 liposomes were intravenously injected and tumor accumulation was evaluated using NIR fluorescent imaging. Western blot and flow cytometry demonstrated that PLX sensitive and resistant A2058 and A375 melanoma cells highly express EMMPRIN. S100A9 liposomes were 200xa0nm diameter and uniformly sized. Flow cytometry demonstrated 100X more intracellular dye uptake by A2058xa0cells treated with S100A9 liposomes compared with untargeted liposomes. Inxa0vivo accumulation of S100A9 liposomes within subcutaneous A2058 and A2058PLX tumors was observed from 6-48xa0h, with A2058PLX accumulating significantly higher levels (Pxa0=xa00.001626). EMMPRIN-targeted liposomes via an S100A9 ligand are a novel, targeted delivery system which could provide improved EMMPRIN specific drug delivery to a tumor.