EMAIL THIS PAGE TO A FRIEND
Neuropeptides
Involvement of hypothalamic neuropeptide Y in pentazocine induced suppression of food intake in rats.
PMID 24656792
Abstract
The potent orexigenic peptide neuropeptide Y (NPY) has been considered as a possible endogenous ligand for a subpopulation of sigma receptors (SigR). However, their mutual interaction with reference to feeding behavior remains poorly understood. In the present study, we explored the possible interaction between sigma1 receptors (Sig1R) agonist, pentazocine, and NPY on food intake in satiated rats. While pentazocine dose-dependently reduced the food intake, NPY significantly increased it at 2, 4 and 6h post injection time points. In combination studies, pretreatment with NPY (0.1 nmol/rat, intra-PVN) normalized the inhibitory effect of pentazocine (60 μg/rat, intra-PVN) on food intake. Similarly, pre-treatment with pentazocine (30 μg/rat, intra-PVN) significantly antagonized the orexigenic effect of NPY (0.5 and 1.0 nmol/rat, intra-PVN). Moreover, pentazocine treatment decreased NPY immunoreactivity in arcuate (ARC), paraventricular (PVN), dorsomedial (DMH) and ventromedial (VMH) nuclei of hypothalamus. However, no change was observed in lateral hypothalamus (LH). Study implicates the reduced NPY immunoreactivity for the anorectic effect observed following pentazocine injections. Therefore, the concomitant activation of the NPYergic system along with the Sig1R agonist treatment may serve a useful purpose in the management of the unwanted side effects related to energy homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P127
(+)-Pentazocine, solid
C19H27NO
P134
(−)-Pentazocine, ≥98% (HPLC)
C19H27NO