EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Simultaneous determination of various bioactive redox components in Shuang-Huang-Lian preparations using a novel three-channel isocratic elution liquid chromatography with electrochemical detection system.
PMID 24657677
Abstract
A novel three-channel isocratic elution high performance liquid chromatography with electrochemical detection (3LC-ECD) system was developed for the simultaneous determination of various bioactive redox components in Shuang-Huang-Lian (SHL) preparations. This 3LC-ECD system consists of three isocratic elution flow ways, two switching valves, four columns, and three electrochemical detectors. Through alternately rotating switching valves to change the elution flow way, six caffeoylquinic acids, four flavonoids, and one phenylethanoid glycoside compound were simultaneously determined within 70 min by the present 3LC-ECD method, in which three compounds of protocatechuic aldehyde, propyl gallate, and kaempferol were used as internal standards. The results demonstrated that the present 3LC-ECD method has achieved desired linearity (r>0.999), precision (relative standard deviation <2.5%), accuracy (recovery, 95.6-103.6%), and high sensitivity (limit of detection, 0.11-0.90 ng/mL). In addition, we successfully determined the contents of these 11 bioactive redox components in 14 batches of SHL oral liquid and 12 batches of SHL lyophilized powder for injection produced by different manufacturers in China.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
65969
4-O-Caffeoylquinic acid, ≥98.0%
C16H18O9
465119
Baicalein, 98%
C15H10O5
572667
Baicalin, 95%
C21H18O11
94121
Baicalin, ≥99.0% (HPLC)
C21H18O11
Y0001273
Baicalin, European Pharmacopoeia (EP) Reference Standard
C21H18O11
C0625
Caffeic acid, ≥98.0% (HPLC)
C9H8O4
60018
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C9H8O4
60020
Caffeic acid, purum, ≥95.0% (HPLC)
C9H8O4
C3878
Chlorogenic acid, ≥95% (titration)
C16H18O9
00500590
Chlorogenic acid, primary pharmaceutical reference standard
C16H18O9
Y0000569
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
C16H18O9
90467
Cryptochlorogenic acid, analytical standard
C16H18O9
96353
Kaempferol, analytical standard
C15H10O6
00550580
Kaempferol, primary pharmaceutical reference standard
C15H10O6
60010
Kaempferol, ≥97.0% (HPLC)
C15H10O6
K0133
Kaempferol, ≥90% (HPLC), powder
C15H10O6
94419
Neochlorogenic acid, from Lonicera japonica, ≥98.0% (HPLC)
C16H18O9
91213
Neochlorogenic acid, analytical standard
C16H18O9
P3130
Propyl gallate, powder
C10H12O5
02370
Propyl gallate, for microscopy, ≥98.0% (HPLC)
C10H12O5
48710
Propyl gallate, antioxidant, ≥98.0% (HPLC)
C10H12O5
W294705
Propyl gallate, ≥98%, FCC
C10H12O5
1576800
Propyl gallate, United States Pharmacopeia (USP) Reference Standard
C10H12O5
P3640000
Propyl gallate, European Pharmacopoeia (EP) Reference Standard
C10H12O5
PHR1118
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O5
73577
Scutellarin, analytical standard
C21H18O12
SMB00081
Wogonoside, ≥95% (LC/MS-ELSD)
C22H20O11