EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Composition of innate lymphoid cell subsets in the human skin: enrichment of NCR(+) ILC3 in lesional skin and blood of psoriasis patients.
PMID 24658504
Abstract
Innate lymphoid cells (ILCs) are increasingly appreciated as important regulators of tissue homeostasis and inflammation. However, their role in human skin remains obscure. We found that healthy peripheral blood CD117(+) ILC3, lacking the natural cytotoxicity receptor (NCR) NKp44 (NCR(-) ILC3), CD117(-)NCR(-)CRTH2(-)CD161(+) ILC1, and CRTH2(+) ILC2, express the skin-homing receptor cutaneous lymphocyte antigen (CLA). NCR(+) ILC3 were scarce in peripheral blood. Consistently, we identified in normal skin ILC2 and NCR(-) ILC3, a small proportion of CD161(+) ILC1, and hardly any NCR(+) ILC3, whereas NCR(+) ILC3 were present in cultured dermal explants. The skin ILC2 and NCR(+) ILC3 subsets produced IL-13 and IL-22, respectively, upon cytokine stimulation. Remarkably, dermal NCR(-) ILC3 converted to NCR(+) ILC3 upon culture in IL-1β plus IL-23, cytokines known to be involved in psoriatic inflammation. In line with this observation, significantly increased proportions of NCR(+) ILC3 were present in lesional skin and peripheral blood of psoriasis patients as compared with skin and blood of healthy individuals, respectively, whereas the proportions of ILC2 and CD161(+) ILC1 remained unchanged. NCR(+) ILC3 from skin and blood of psoriasis patients produced IL-22, which is regarded as a key driver of epidermal thickening, suggesting that NCR(+) ILC3 may participate in psoriasis pathology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9868 Anti-CRTH2 (GPR44) antibody produced in chicken, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500194
Anti-CRTH2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4877
Anti-IL-17 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4887
Anti-IL-17 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2502114
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502115
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA052258
Anti-IL17A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502005
Anti-IL7R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406026
Anti-IL7R antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410710
Anti-IL7R antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300489
Anti-KIT (Ab-721) antibody produced in rabbit, affinity isolated antibody
SAB4300593
Anti-KIT (Ab-936) antibody produced in rabbit, affinity isolated antibody
GW21114 Anti-KIT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412042
ANTI-KIT antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412043
ANTI-KIT antibody produced in mouse, clone 5F6, purified immunoglobulin, buffered aqueous solution
SAB1412044
ANTI-KIT antibody produced in mouse, clone 5A11, purified immunoglobulin, buffered aqueous solution
SAB1412045
ANTI-KIT antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1412046
ANTI-KIT antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412140
ANTI-KIT antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution
SAB1412141
ANTI-KIT antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
HPA004471
Anti-KIT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039113 Anti-KLRB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700234
Anti-Ncam1 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501672
Anti-NCAM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4300660
Anti-NCAM1 antibody produced in rabbit, affinity isolated antibody
SAB3500881
Anti-PTGDR2 antibody produced in rabbit, affinity isolated antibody
SAB4700750
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
SAB4700273
Monoclonal Anti-CD16-PE antibody produced in mouse, clone LNK16, purified immunoglobulin, buffered aqueous solution
SAB4700279
Monoclonal Anti-CD16-PE antibody produced in mouse, clone MEM-154, purified immunoglobulin, buffered aqueous solution
SAB4700689 Monoclonal Anti-CD16-PE antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution