EMAIL THIS PAGE TO A FRIEND
The Analyst
3D-printed fluidic devices enable quantitative evaluation of blood components in modified storage solutions for use in transfusion medicine.
PMID 24660218
Abstract
A fluidic device constructed with a 3D-printer can be used to investigate stored blood components with subsequent high-throughput calibration and readout with a standard plate reader.