EMAIL THIS PAGE TO A FRIEND
Oncogene
Intratumoral heterogeneity of ADAM23 promotes tumor growth and metastasis through LGI4 and nitric oxide signals.
PMID 24662834
Abstract
Intratumoral heterogeneity (ITH) represents an obstacle for cancer diagnosis and treatment, but little is known about its functional role in cancer progression. The A Desintegrin And Metalloproteinase 23 (ADAM23) gene is epigenetically silenced in different types of tumors, and silencing is often associated with advanced disease and metastasis. Here, we show that invasive breast tumors exhibit significant ADAM23-ITH and that this heterogeneity is critical for tumor growth and metastasis. We demonstrate that while loss of ADAM23 expression enhances invasion, it causes a severe proliferative deficiency and is not itself sufficient to trigger metastasis. Rather, we observed that, in ADAM23-heterotypic environments, ADAM23-negative cells promote tumor growth and metastasis by enhancing the proliferation and invasion of adjacent A23-positive cells through the production ofxa0LGI4 (Leucine-rich Glioma Inactivated 4) and nitric oxide (NO). Ablation of LGI4 and NO in A23-negative cells significantly attenuates A23-positive cell proliferation and invasion. Our work denotes a driving role of ADAM23-ITH during disease progression, shifting the malignant phenotype from the cellular to the tissue level. Our findings also provide insights for therapeutic intervention, enforcing the need to ascertain ITH to improve cancer diagnosis and therapy.