EMAIL THIS PAGE TO A FRIEND
Circulation
Coinhibitory suppression of T cell activation by CD40 protects against obesity and adipose tissue inflammation in mice.
PMID 24664276
Abstract
Costimulatory cascades such as the CD40L-CD40 dyad enhance immune cell activation and inflammation during atherosclerosis. Here, we tested the hypothesis that CD40 directly modulates traits of the metabolic syndrome in diet-induced obesity in mice. To induce the metabolic syndrome, wild-type or CD40(-/-) mice consumed a high-fat diet for 20 weeks. Unexpectedly, CD40(-/-) mice exhibited increased weight gain, impaired insulin secretion, augmented accumulation of inflammatory cells in adipose tissue, and enhanced proinflammatory gene expression. This proinflammatory and adverse metabolic phenotype could be transplanted into wild-type mice by reconstitution with CD40-deficient lymphocytes, indicating a major role for CD40 in T or B cells in this context. Conversely, therapeutic activation of CD40 signaling by the stimulating antibody FGK45 abolished further weight gain during the study, lowered glucose levels, improved insulin sensitivity, and suppressed adipose tissue inflammation. Mechanistically, CD40 activation decreased the expression of proinflammatory cytokines in T cells but not in B cells or macrophages. Finally, repopulation of lymphocyte-free Rag1(-/-) mice with CD40(-/-) T cells provoked dysmetabolism and inflammation, corroborating a protective role of CD40 on T cells in the metabolic syndrome. Finally, levels of soluble CD40 showed a positive association with obesity in humans, suggesting clinical relevance of our findings. We present the surprising finding that CD40 deficiency on T cells aggravates whereas activation of CD40 signaling improves adipose tissue inflammation and its metabolic complications. Therefore, positive modulation of the CD40 pathway might describe a novel therapeutic concept against cardiometabolic disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41929
Anti-CD40 antibody produced in rabbit, affinity isolated antibody
HPA031567
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031568
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103449
Anti-CD40, (N-terminal) antibody produced in rabbit, affinity isolated antibody
RAB0085
Human CD40 ELISA Kit, for serum, plasma, cell culture supernatant and urine
SAB4700268 Monoclonal Anti-CD16 antibody produced in mouse, clone MEM-168, purified immunoglobulin, buffered aqueous solution
SAB4700269 Monoclonal Anti-CD16 antibody produced in mouse, clone LNK16, purified immunoglobulin, buffered aqueous solution
SAB4700274 Monoclonal Anti-CD16 antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution
SAB4700276 Monoclonal Anti-CD16 antibody produced in mouse, clone MEM-154, purified immunoglobulin, buffered aqueous solution
WH0000958M1
Monoclonal Anti-CD40 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
AMAB90905
Monoclonal Anti-CD40 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1673, purified immunoglobulin, buffered aqueous glycerol solution
RAB0086
Mouse CD40 ELISA Kit, for serum, plasma and cell culture supernatant