EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Postprandial triglyceride-rich lipoproteins promote invasion of human coronary artery smooth muscle cells in a fatty-acid manner through PI3k-Rac1-JNK signaling.
PMID 24668798
Abstract
The aim was to investigate the effect of postprandial triglyceride-rich lipoproteins (TRLs) with different fatty acid compositions on human coronary artery smooth muscle cell (hCASMC) invasion and to identify the molecular pathways involved. TRLs were isolated from the plasma of healthy volunteers after the ingestion of single meals enriched in MUFAs, saturated fatty acids (SFAs), or PUFAs. hCASMC invasion was analyzed using transwell chambers with Matrigel. TRLs-SFAs provoked the highest invasion, followed by TRLs-MUFAs and TRLs-PUFAs. Inhibition studies with Orlistat showed that invasion was dependent on the fatty acid composition of the TRLs. Fatty acids incorporated into the cell membranes strongly associated with cell invasion. Pull-down assays showed that TRLs-SFAs were able to increase Rac1 activity via inhibition of RhoA-dependent signaling. Chemical inhibition and siRNA studies showed that Rac1, PI3k, JNK, and MMP2 regulates TRL-SFA-induced hCASMC invasion. We demonstrate for the first time that TRLs induce hCASMCs invasion in a fatty acid dependent manner. This effect in TRLs-SFAs is mediated by the PI3k-Rac1-JNK, RhoA, and Rac1-MMP2 pathways. The ingestion of MUFA, compared to other dietary fatty acids such as SFA, could be considered as a nutritional strategy to reduce the atherosclerotic plaque formation.