EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Alpha 2-macroglobulin.
PMID 2467173
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63013
α2-Macroglobulin from human plasma, ≥90% (GE)