EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Lipid-lipid interactions in aminated reduced graphene oxide interface for biosensing application.
PMID 24673363
Abstract
A label-free biosensor based on antiapolipoprotein B 100 functionalized-aminated reduced graphene oxide interface has been fabricated for detection of low density lipoprotein (LDL or lipid) cholesterol. The aminated reduced graphene oxide (NH2-rGO) based electrode surface is covalently functionalized with antiapolipoprotein B 100 (AAB or lipid) using EDC/NHS coupling chemistry. The lipid-lipid interactions at the NH2-rGO electrode surface have been investigated using electrochemical impedance spectroscopic technique. The structural and morphological investigations of NH2-rGO based immunosensor have been accomplished via transmission electron microscopy, X-ray diffraction, Fourier transform infrared spectroscopy, UV-visible, and electrochemical techniques. The impedimetric response of the proposed immunosensor shows excellent sensitivity (612 Ω mg(-1) dL cm(-2)), a response time of 250 s, and a low detection limit of 5 mg/dL of LDL molecules. The association, dissociation, and equilibrium rate constants for this immunoelectrode are found to be 1.66 M(-1) s(-1), 0.6 s(-1), and 2.77 M(-1), respectively. The long-term stability and excellent reproducibility of the proposed immunosensor indicates a suitable platform for detection of LDL or lipid molecules. This immunosensor provides an efficient platform for analysis of the antigen-antibody interactions of lipid molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
805432
Reduced graphene oxide, amine functionalized