EMAIL THIS PAGE TO A FRIEND
Arthritis & Rheumatology (Hoboken, N.J.)
A122: metabolomic analysis of breath volatile organic compounds reveals unique breathprints in children with juvenile idiopathic arthritis.
PMID 24677875
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30650
1-Decene, ≥97.0% (GC)
C10H20