EMAIL THIS PAGE TO A FRIEND
Pediatric and developmental pathology : the official journal of the Society for Pediatric Pathology and the Paediatric Pathology Society
Skin of patients with large/giant congenital melanocytic nevi shows increased mast cells.
PMID 24679055
Abstract
Nevocytes (NC) and mastocytes (MC) have different progenitors but share stem cell factor as regulator/activator of NC and for differentiation/proliferation of MC. Both cell types express stem cell factor receptor CD117. We hypothesize that large/giant congenital melanocytic nevi (L/GCMN) may associate with MC hyperplasia. Forty-nine L/GCMN were examined, 12 samples from uninvolved skin of L/GCMN patients and 6 control skin samples studied with Giemsa and immunohistochemistry for CD117 and MC-tryptase. Picrosirius red (PR) was used to assess fibrosis. Digital images were used to count MC/mm(2) using ImageJ software. Western blot (WB) for MC-tryptase in 12 GCMN and 12 non-nevus samples was performed. Analysis of variance (Tukey) and Pearson statistical tests were applied. Increased MCs were observed in nevus tissue (75.1 ± 35.3 MCs/mm(2)) and in uninvolved skin (53.74 ± 27.7 MC/ mm(2)). P  =  0.109 from patients with L/GCMN, compared with controls from individuals without L/GCMN (28.74 ± 8.4 MC/mm(2)); P  =  0.001 supported by results of WB analysis for tryptase. A positive trend toward correlation of MC numbers with fibrosis, assessed by PR staining fell short of statistical significance (r  =  0.245; P  =  0.086); no difference in fibrosis was found between nevus and non-nevus skin from patients with L/GCMN (P  =  0.136). We found a higher density of MC, both in normal-appearing skin and nevus areas of L/GCMN patients, compared with control skin samples from individuals without nevi. Given the abnormal wound healing and allergic reactions described in L/GCMN patients, these findings suggest a potential role for MC in the biology of L/GCMN, making them a potential target for therapeutic intervention.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300489
Anti-KIT (Ab-721) antibody produced in rabbit, affinity isolated antibody
SAB4300593
Anti-KIT (Ab-936) antibody produced in rabbit, affinity isolated antibody
GW21114 Anti-KIT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412042
ANTI-KIT antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412043
ANTI-KIT antibody produced in mouse, clone 5F6, purified immunoglobulin, buffered aqueous solution
SAB1412044
ANTI-KIT antibody produced in mouse, clone 5A11, purified immunoglobulin, buffered aqueous solution
SAB1412045
ANTI-KIT antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1412046
ANTI-KIT antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412140
ANTI-KIT antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution
SAB1412141
ANTI-KIT antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
HPA004471
Anti-KIT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1303861
ANTI-TPSAB1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1409765
Anti-TPSAB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411362
Anti-TPSAB1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA049153
Anti-TPSB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700750
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
WH0003815M2
Monoclonal Anti-KIT antibody produced in mouse, clone 6F2, purified immunoglobulin, buffered aqueous solution
AMAB90901
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1657, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90904
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1667, purified immunoglobulin, buffered aqueous glycerol solution
SAB1404007
Monoclonal Anti-KIT antibody produced in mouse, clone 6G12, purified immunoglobulin, buffered aqueous solution
SAB5300439
Monoclonal Anti-KIT antibody produced in mouse, clone 8D7, ascites fluid
SAB5300440
Monoclonal Anti-KIT antibody produced in mouse, clone 1C5, ascites fluid
WH0007177M1
Monoclonal Anti-TPSAB1 antibody produced in mouse, clone 2A10-B5, purified immunoglobulin, buffered aqueous solution