EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
A eukaryotic-like 3' untranslated region in Salmonella enterica hilD mRNA.
PMID 24682814
Abstract
Long 3' untranslated regions (3'UTRs) are common in eukaryotic mRNAs. In contrast, long 3'UTRs are rare in bacteria, and have not been characterized in detail. We describe a 3'UTR of 310 nucleotides in hilD mRNA, a transcript that encodes a transcriptional activator of Salmonella enterica pathogenicity island 1 (SPI-1). Deletion of the hilD 3'UTR increases the hilD mRNA level, suggesting that the hilD 3'UTR may play a role in hilD mRNA turnover. Cloning of the hilD 3'UTR downstream of the green fluorescent protein (gfp) gene decreases green fluorescent protein (GFP) activity in both Escherichia coli and S. enterica, indicating that the hilD 3'UTR can act as an independent module. S. enterica mutants lacking either ribonuclease E or polynucleotide phosphorylase contain similar amounts of hilD and hilD Δ3'UTR mRNAs, suggesting that the hilD 3'UTR is a target for hilD mRNA degradation by the degradosome. The hilD 3'UTR is also necessary for modulation of hilD and SPI-1 expression by the RNA chaperone Hfq. Overexpression of SPI-1 in the absence of the hilD 3'UTR retards Salmonella growth and causes uncontrolled invasion of epithelial cells. Based on these observations, we propose that the S. enterica hilD 3'UTR is a cis-acting element that contributes to cellular homeostasis by promoting hilD mRNA turnover.