EMAIL THIS PAGE TO A FRIEND
Clinical and experimental pharmacology & physiology
Oestrogen upregulates the sarcoplasmic reticulum Ca(2+) ATPase pump in coronary arteries.
PMID 24684418
Abstract
The presence of circulating plasma 17β-oestradiol (E2) is beneficial in women against abnormal vascular tone development, such as coronary arterial vasospasms. Several vascular diseases have demonstrated that increased expression of the sarcoplasmic reticulum Ca(2+) -ATPase pump (SERCA2b) serves to limit the excessive accumulation of intracellular Ca(2+) . Therefore, the hypothesis of the present study was that E2 would increase SERCA2b expression in the coronary vasculature. Coronary arteries were dissected from hearts obtained from mature female pigs. Artery segments were cultured for 24xa0h in E2 (1xa0pmol/L or 1xa0nmol/L) and homogenized for western blot analysis. At 1xa0nmol/L, E2 induced an approximate 50% increase in immunoreactivity for SERCA2b. In addition, E2 increased the protein expression of the known SERCA regulatory proteins, protein kinase A (PKA) and protein kinase G (PKG). The E2-induced increase in SERCA2b was attenuated when the culture medium was supplemented with the oestrogen receptor (ER) α/β antagonist ICI 182,780 and the PKG antagonist KT5823 (10xa0μmol/L, 24xa0h for both). The PKA antagonist (KT5720; 10xa0μmol/L, 24xa0h) had no effect on SERCA2b expression. Removal of the endothelium (using a wooden toothpick) from artery segments prior to culture decreased the E2-mediated increase in SERCA2b and PKG expression by 45% and 47%, respectively. Overall, the findings suggest that one of the potential cardiovascular benefits of E2 in women is upregulation of SERCA2b, via activation of the classic ERα and ERβ pathway.