EMAIL THIS PAGE TO A FRIEND
Cancer research
LEO1 is regulated by PRL-3 and mediates its oncogenic properties in acute myelogenous leukemia.
PMID 24686170
Abstract
PRL-3, an oncogenic dual-specificity phosphatase, is overexpressed in 50% of acute myelogenous leukemia (AML) and associated with poor survival. We found that stable expression of PRL-3 confers cytokine independence and growth advantage of AML cells. However, how PRL-3 mediates these functions in AML is not known. To comprehensively screen for PRL3-regulated proteins in AML, we performed SILAC-based quantitative proteomics analysis and discovered 398 significantly perturbed proteins after PRL-3 overexpression. We show that Leo1, a component of RNA polymerase II-associated factor (PAF) complex, is a novel and important mediator of PRL-3 oncogenic activities in AML. We described a novel mechanism where elevated PRL-3 protein increases JMJD2C histone demethylase occupancy on Leo1 promoter, thereby reducing the H3K9me3 repressive signals and promoting Leo1 gene expression. Furthermore, PRL-3 and Leo1 levels were positively associated in AML patient samples (N=24; P<0.01). On the other hand, inhibition of Leo1 reverses PRL-3 oncogenic phenotypes in AML. Loss of Leo1 leads to destabilization of the PAF complex and downregulation of SOX2 and SOX4, potent oncogenes in myeloid transformation. In conclusion, we identify an important and novel mechanism by which PRL-3 mediates its oncogenic function in AML.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2700513
Anti-CTR9 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300412
Anti-HDAC2 (Ab-394) antibody produced in rabbit, affinity isolated antibody
HPA011727
Anti-HDAC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32618
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB2104633
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
H3159
Anti-Histone Deacetylase 2 (HDAC2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H2663
Anti-Histone Deacetylase 2 (HDAC2) antibody, Mouse monoclonal, clone HDAC2-62, purified from hybridoma cell culture
SAB2101192
Anti-JMJD2C antibody produced in rabbit, affinity isolated antibody
SAB1300629
Anti-LEO1 (S551) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA040741
Anti-LEO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA040941
Anti-LEO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106984
Anti-LEO1 antibody produced in rabbit, affinity isolated antibody
SAB2700050
Anti-PAF1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700103
Anti-PAF1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA041875
Anti-PAF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107634
Anti-PAF1 antibody produced in rabbit, affinity isolated antibody
SAB2107635
Anti-PAF1 antibody produced in rabbit, affinity isolated antibody
P0498 Anti-PRL-3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306065
ANTI-PRL3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
GW22107F Anti-PTP4A3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2701818
Anti-ptp4a3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101908
Anti-PTP4A3 antibody produced in rabbit, affinity isolated antibody
SAB4300569
Anti-PTP4A3 antibody produced in rabbit, affinity isolated antibody
SAB1407065
Anti-SMR3B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2106583
Anti-SMR3B antibody produced in rabbit, affinity isolated antibody
A1270000
Arginine, European Pharmacopoeia (EP) Reference Standard
C6H14N4O2
11020
DL-Arginine, ≥97.0% (NT)
C6H14N4O2
SAB1305533 MONOCLONAL ANTI-PRL3 antibody produced in mouse, clone 111AT714, IgG fraction of antiserum, buffered aqueous solution