EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
The effect of Lfcin-B on non-small cell lung cancer H460 cells is mediated by inhibiting VEGF expression and inducing apoptosis.
PMID 24691828
Abstract
Lfcin-B, an antimicrobial peptide found in various exocrine secretions of mammals, showed antitumor effects. However, the effect and relative mechanism of Lfcin-B on non-small cell lung cancer is unclear. In this study, assay of cell viability, quantitative real-time PCR, Western blot, annexin V/propidium iodide assay, flow cytometry and tumor-xenograft model were applied to elucidate the mechanism of Lfcin-B on non-small cell lung cancer NCI-H460 (H460) cells. Lfcin-B significantly suppressed the proliferation of H460 cells in vitro. Additionally, the transcription and translation of the VEGF gene in H460 cells were restrained after exposure to Lfcin-B. Moreover, the apoptosis of H460 cells was induced by Lfcin-B through stimulating caspase-3, caspase-9 and preventing survivin expression on both the transcription and translation level. Meanwhile, Lfcin-B increased the production of reactive oxygen species and suppressed the RNA of antioxidant enzymes (GPX1, GPX2, SOD3 and catalase) in H460 cells. Finally, Lfcin-B significantly prevented the tumor growth in the H460-bearing mice model. These results indicated that Lfcin-B could be a potential candidate for the treatment of lung cancer.