EMAIL THIS PAGE TO A FRIEND
Cell biochemistry and biophysics
Toxicity of copper on isolated liver mitochondria: impairment at complexes I, II, and IV leads to increased ROS production.
PMID 24691927
Abstract
Oxidative damage has been implicated in disorders associated with abnormal copper metabolism and also Cu(2+) overloading states. Besides, mitochondria are one of the most important targets for Cu(2+), an essential redox transition metal, induced hepatotoxicity. In this study, we aimed to investigate the mitochondrial toxicity mechanisms on isolated rat liver mitochondria. Rat liver mitochondria in both in vivo and in vitro experiments were obtained by differential ultracentrifugation and the isolated liver mitochondria were then incubated with different concentrations of Cu(2+). Our results showed that Cu(2+) induced a concentration and time-dependent rise in mitochondrial ROS formation, lipid peroxidation, and mitochondrial membrane potential collapse before mitochondrial swelling ensued. Increased disturbance in oxidative phosphorylation was also shown by decreased ATP concentration and decreased ATP/ADP ratio in Cu(2+)-treated isolated mitochondria. In addition, collapse of mitochondrial membrane potential (MMP), mitochondrial swelling, and release of cytochrome c following of Cu(2+) treatment were well inhibited by pretreatment of mitochondria with CsA and BHT. Our results showed that Cu(2+) could interact with respiratory complexes (I, II, and IV). This suggests that Cu(2+)-induced liver toxicity is the result of metal's disruptive effect on liver hepatocyte mitochondrial respiratory chain that is the obvious cause of Cu(2+)-induced ROS formation, lipid peroxidation, mitochondrial membrane potential decline, and cytochrome c expulsion which start cell death signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19422
1-Butanol, analytical standard
C4H10O
154679
1-Butanol, spectrophotometric grade, 99.5%
C4H10O
19420
1-Butanol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C4H10O
442665
1-Butanol, analytical standard
C4H10O
PHR1209
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
34750
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
C15H24O
03762
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph.Eur.
C15H24O
1082708
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
C15H24O
PHR1117
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
C15H24O
C3662
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
C62H111N11O12
30024
Cyclosporin A, ≥98.5% (TLC)
C62H111N11O12
32425
Cyclosporin A, VETRANAL, analytical standard
C62H111N11O12
PHR1092
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
C62H111N11O12
W327700
Disodium succinate, ≥98%
C4H4Na2O4
00489
Ethylene, puriss., ≥99.95% (GC)
C2H4
03482
Ethylene, purum, ≥99.9%
C2H4
03484
Ethylene, ≥99.9%
C2H4
H1252
Homovanillic acid, Fluorimetric reagent
C9H10O4
69673
Homovanillic acid, analytical standard
C9H10O4
14160
Sodium succinate dibasic, purum, anhydrous, ≥98.0% (NT)
C4H4Na2O4