EMAIL THIS PAGE TO A FRIEND
Journal of virology
Human papillomavirus type 16 E6* induces oxidative stress and DNA damage.
PMID 24696478
Abstract
High-risk types of human papillomavirus (HPV) are the causative agents of virtually all cases of cervical cancer and a significant proportion of other anogenital cancers, as well as both oral and pharyngeal cancers. The high-risk types encode two viral oncogenes, E6 and E7, which work together to initiate cell transformation. Multiple steps involving the activities and interactions of both viral and cellular proteins are involved in the progression from HPV infection to cell transformation to cancer. The E6 oncoprotein is expressed as several isoforms: a full-length variant referred to as E6 and a few shorter isoforms collectively referred to as E6*. In this study, we found that expression of E6* increased the level of reactive oxygen species (ROS) in both HPV-positive and HPV-negative cells. This increased oxidative stress led to higher levels of DNA damage, as assessed by the comet assay, quantification of 8-oxoguanine, and poly(ADP-ribose) polymerase 1. The observed increase in ROS may be due to a decrease in cellular antioxidant activity, as we found that E6* expression also led to decreased expression of superoxide dismutase isoform 2 and glutathione peroxidase. These studies indicate that E6* may play an important role in virus-induced mutagenesis by increasing oxidative stress and DNA damage. Our findings demonstrate for the first time that an HPV gene product, E6*, can increase ROS levels in host cells. This ability may play a significant role both in the viral life cycle and in cancer development, because an increase in oxidative DNA damage may both facilitate HPV genome amplification and increase the probability of HPV16 DNA integration. Integration, in turn, is thought to be an important step in HPV-mediated carcinogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500468
Anti-Glutathione peroxidase 1 (ISO1) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502102
Anti-Gpx1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502103
Anti-Gpx1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2700534
Anti-GPX1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA044758
Anti-GPX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500976
Anti-SOD1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406464
Anti-SOD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001401
Anti-SOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV45752
Anti-SOD1 antibody produced in rabbit, IgG fraction of antiserum
SAB5200083
Anti-SOD1 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
SAB1411305
Anti-SOD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501675
Anti-SOD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501676
Anti-SOD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406465
Anti-SOD2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701618
Anti-sod2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701761
Anti-sod2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001814
Anti-SOD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102261
Anti-SOD2 antibody produced in rabbit, affinity isolated antibody
EHU075891 MISSION® esiRNA, esiRNA human SOD2 (esiRNA1)
EMU077671 MISSION® esiRNA, esiRNA targeting mouse Sod2 (esiRNA1)
SAB1409700
Monoclonal Anti-SOD1 antibody produced in mouse, clone 10D5, purified immunoglobulin, buffered aqueous solution