EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Functional understanding of the versatile protein copper metabolism MURR1 domain 1 (COMMD1) in copper homeostasis.
PMID 24697840
Abstract
Copper is an important cofactor in numerous biological processes in all living organisms. However, excessive copper can be extremely toxic, so it is vital that the copper level within a cell is tightly regulated. The damaging effect of copper is seen in several hereditary forms of copper toxicity in humans and animals. At present, Wilson's disease is the best-described and best-studied copper-storage disorder in humans; it is caused by mutations in the ATP7B gene. In dogs, a mutation in the COMMD1 gene has been found to be associated with copper toxicosis. Using a liver-specific Commd1 knockout mouse, the biological role of Commd1 in copper homeostasis has been confirmed. Yet, the exact mechanism by which COMMD1 regulates copper homeostasis is still unknown. Here, we give an overview of the current knowledge and perspectives on the molecular function of COMMD1 in copper homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF71667366
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Cu
GF21709650
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF08494577
Copper, wire reel, 1000m, diameter 0.075mm, annealed, 99.9%
Cu
GF66965956
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Cu
GF22319872
Copper, wire reel, 1000m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF27350287
Copper, wire reel, 1000m, diameter 0.1mm, annealed, 99.9%
Cu
GF63655780
Copper, wire reel, 1000m, diameter 0.25mm, annealed, 99.9%
Cu
GF47171033
Copper, wire reel, 100m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF18312791
Copper, wire reel, 100m, diameter 0.02mm, hard, 99.99+%
Cu
GF89712333
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Cu
GF93417105
Copper, wire reel, 100m, diameter 1.0mm, as drawn, 99.98+%
Cu
GF64298829
Copper, wire reel, 100m, diameter 0.02mm, annealed, 99.99%
Cu
GF86528155
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Cu
GF39045090
Copper, wire reel, 100m, diameter 0.075mm, annealed, 99.9%
Cu
GF71409089
Copper, wire reel, 100m, diameter 0.125mm, annealed, 99.9%
Cu
GF42860651
Copper, wire reel, 100m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF52128008
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Cu
GF69188079
Copper, wire reel, 100m, diameter 0.25mm, annealed, 99.9%
Cu
GF98066054
Copper, wire reel, 100m, diameter 0.5mm, annealed, 99.9%
Cu
GF81594399
Copper, wire reel, 100m, diameter 0.5mm, annealed, silver plated, 99.9%
Cu
GF82471620
Copper, wire reel, 100m, diameter 1.0mm, annealed, 99.9%
Cu
GF83100619
Copper, wire reel, 10m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF80026770
Copper, wire reel, 10m, diameter 0.02mm, hard, 99.99+%
Cu
GF74431324
Copper, wire reel, 10m, diameter 0.05mm, as drawn, 99.99+%
Cu
GF81301609
Copper, wire reel, 10m, diameter 0.75mm, as drawn, 99.98+%
Cu
GF86675439
Copper, wire reel, 10m, diameter 0.01mm, hard, 99.99%
Cu
GF77356109
Copper, wire reel, 10m, diameter 0.02mm, annealed, 99.99%
Cu
GF56973257
Copper, wire reel, 2000m, diameter 0.25mm, annealed, 99.9%
Cu