EMAIL THIS PAGE TO A FRIEND
Basic & clinical pharmacology & toxicology
Puerarin attenuates carbon tetrachloride-induced liver oxidative stress and hyperlipidaemia in mouse by JNK/c-Jun/CYP7A1 pathway.
PMID 24698568
Abstract
Puerarin (PU), a natural flavonoid, has been reported to have many benefits and medicinal properties. The aim of this study was to investigate the effects of puerarin on hepatic oxidative stress and hyperlipidaemia in mice exposed to carbon tetrachloride (CCl4). Male ICR mice were injected with CCl4 with or without puerarin co-administration (200 and 400xa0mg/kg intragastrically once-daily) for 8xa0weeks. Our data showed that puerarin significantly prevented CCl4-induced hepatotoxicity, indicated by both diagnostic indicators of the liver damage (serum aminotransferase levels) and histopathological analysis. Puerarin decreased the thiobarbituric acid reactive substances (TBARS) and the protein carbonyl content (PCO) in the liver of CCl4-treated mice. Puerarin also restored the levels of reduced glutathione (GSH) and total antioxidant capacity (TAC) in the liver. Furthermore, the increase in serum cholesterol, triglycerides and low-density lipoproteins (LDL) induced by CCl4 was effectively suppressed by puerarin. The high-density lipoprotein (HDL) level in the CCl4 treatment mice was also increased by puerarin. Western blot analysis showed that puerarin remarkably inhibited hyperlipidaemia by regulating the expression of phosphorylated Jun N-terminal kinases (JNK), phosphorylated c-Jun protein and cholesterol 7a-hydroxylase (CYP7A1) in the liver of CCl4-treated mice. Altogether, these results suggest that puerarin could protect the CCl4-induced liver injury and hyperlipidaemia by reducing reactive oxygen species S production, renewing the total antioxidant capacity and influencing expression of hepatic lipid biosynthesis and metabolism genes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4504451
Anti-phospho-c-Jun (pSer243) antibody produced in rabbit, affinity isolated antibody
J2128 Anti-phospho-c-Jun (pSer63) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504368
Anti-phospho-c-Jun (pSer63) antibody produced in rabbit, affinity isolated antibody
J2253 Anti-phospho-c-Jun (pSer73) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504369
Anti-phospho-c-Jun (pSer73) antibody produced in rabbit, affinity isolated antibody
SAB4504452
Anti-phospho-c-Jun (pThr239) antibody produced in rabbit, affinity isolated antibody
SAB4504453
Anti-phospho-c-Jun (pThr91) antibody produced in rabbit, affinity isolated antibody
SAB4504454
Anti-phospho-c-Jun (pThr93) antibody produced in rabbit, affinity isolated antibody
SAB4504455
Anti-phospho-c-Jun (pTyr170) antibody produced in rabbit, affinity isolated antibody
SAB4300015
Anti-phospho-JUN (pSer243) antibody produced in rabbit, affinity isolated antibody
SAB4300002
Anti-phospho-JUN (pSer73) antibody produced in rabbit, affinity isolated antibody
SAB4300014
Anti-phospho-JUN (pThr239) antibody produced in rabbit, affinity isolated antibody
SAB4300011
Anti-phospho-JUN (pThr91) antibody produced in rabbit, affinity isolated antibody
SAB4300012
Anti-phospho-JUN (pThr93) antibody produced in rabbit, affinity isolated antibody
SAB4300013
Anti-phospho-JUN (pTyr170) antibody produced in rabbit, affinity isolated antibody
J4399 Monoclonal Anti-phospho-c-Jun (pThr91,93) antibody produced in mouse, ~2 mg/mL, clone C-J 4C4/1, purified immunoglobulin, buffered aqueous solution