EMAIL THIS PAGE TO A FRIEND
Leukemia
Heparanase enhances myeloma progression via CXCL10 downregulation.
PMID 24699306
Abstract
In order to explore the mechanism(s) underlying the pro-tumorigenic capacity of heparanase, we established an inducible Tet-on system. Heparanase expression was markedly increased following addition of doxycycline (Dox) to the culture medium of CAG human myeloma cells infected with the inducible heparanase gene construct, resulting in increased colony number and size in soft agar. Moreover, tumor xenografts produced by CAG-heparanase cells were markedly increased in mice supplemented with Dox in their drinking water compared with control mice maintained without Dox. Consistently, we found that heparanase induction is associated with decreased levels of CXCL10, suggesting that this chemokine exerts tumor-suppressor properties in myeloma. Indeed, recombinant CXCL10 attenuated the proliferation of CAG, U266 and RPMI-8266 myeloma cells. Similarly, CXCL10 attenuated the proliferation of human umbilical vein endothelial cells, implying that CXCL10 exhibits anti-angiogenic capacity. Strikingly, development of tumor xenografts produced by CAG-heparanase cells overexpressing CXCL10 was markedly reduced compared with control cells. Moreover, tumor growth was significantly attenuated in mice inoculated with human or mouse myeloma cells and treated with CXCL10-Ig fusion protein, indicating that CXCL10 functions as a potent anti-myeloma cytokine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21037 Anti-CXCL10 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1410717
Anti-CXCL10 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C1366 Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501241
Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody
SAB3501088
Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody
SAB1405763
Anti-EPHB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4500732
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500775
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500776
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB1412026
ANTI-EPHB2 antibody produced in mouse, clone 1F10, ascites fluid, buffered aqueous solution
SAB2107575
Anti-HPSE antibody produced in rabbit, affinity isolated antibody
SAB2107576
Anti-HPSE antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4300034
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
SAB4504540
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
S2690 Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300033
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
SAB4504541
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
HPA006185
Anti-SDC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300433
Anti-SRC (Ab-529) antibody produced in rabbit, affinity isolated antibody
SAB2102295
Anti-SRC antibody produced in rabbit, affinity isolated antibody
SAB4502847
Anti-SRC antibody produced in rabbit, affinity isolated antibody
SAB2103351
Anti-SRC, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1306786
ANTI-SRC1 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1300410
Anti-SRC1 (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1306922
ANTI-SRC1(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C8490 CRG-2 from mouse, BioReagent, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), lyophilized powder, suitable for cell culture
SRP3246 IP-10 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
SAB1305543 MONOCLONAL ANTI-CD138 antibody produced in mouse, clone 480CT5.4.3, IgG fraction of antiserum, buffered aqueous solution
SAB1305487
MONOCLONAL ANTI-CD138 antibody produced in mouse, clone 480CT5.4.3, crude ascites, buffered aqueous solution
C6473 Monoclonal Anti-CXCR3 antibody produced in mouse, clone 49801.111, purified immunoglobulin, lyophilized powder