EMAIL THIS PAGE TO A FRIEND
European journal of nutrition
Human milk composition differs in healthy mothers and mothers with celiac disease.
PMID 24700375
Abstract
To investigate whether breast-milk composition and microbiota differ in healthy mothers and mothers with celiac disease (CD) to ultimately contribute to identify additional factors determining CD risk. Breast-milk samples from healthy mothers (n = 12) and mothers with CD (n = 12) were collected. Cytokines and secretory immunoglobulin A (sIgA) were analyzed by bead-arrays and flow cytometry and human milk oligosaccharides (HMOs) were assessed by capillary electrophoresis with laser-induced fluorescence (CE-LIF) detection. Breast-milk microbiota composition was analyzed by conventional and quantitative real-time PCR. Breast milk from CD mothers showed significantly lower levels of interleukin (IL) 12p70 (P < 0.042), transforming growth factor (TGF)-β1 (P < 0.018) and sIgA (P < 0.003) and almost significantly lower levels of interferon (IFN)-γ (P < 0.058). Six mothers in each group belonged to the secretor Le(a-b+) type, one to the secretor Le(a-b-) type and five to the non-secretor Le(a+b-) type. CD mothers of non-secretor Le(a+b-) type showed increased Lacto-N-tetraose content (P < 0.042) compared with healthy mothers. CD mothers' milk showed reduced gene copy numbers of Bifidobacterium spp. (P < 0.026) and B. fragilis group (P < 0.044). CD mothers' breast milk is characterized by a reduced abundance of immunoprotective compounds (TGF-β1 and sIgA) and bifidobacteria. The reduction in these components could theoretically diminish the protective effects of breast-feeding on the child's future risk of developing CD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29238
Activated Charcoal Norit®, Norit RBAA-3, rod
C
GF93740384
Carbon - Vitreous, rod, 100mm, diameter 10mm, glassy carbon
C
GF65427587
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
C
GF05962765
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
C
GF68195403
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
C
GF23723423
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
C
GF91785134
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
C
GF78447707
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
C
GF45345562
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
C
GF24425805
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
C
GF93251011
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
C
GF20176680
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
C
GF59400233
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF39088648
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF07378476
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF39873369
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF61342220
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF58042324
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF12408713
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF04031132
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF06230922
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF95357526
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
C
GF61808889
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
C
GF09325009
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
C
GF22691426
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
C
GF74135628
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
C
GF99431523
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
C
GF44361457
Carbon - Vitreous, foil, 200x200mm, thickness 1.0mm, glassy carbon
C
GF10355958
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
C
GF89520521
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
C