EMAIL THIS PAGE TO A FRIEND
Vascular pharmacology
Cyclopiazonic acid alters serotonin-induced responses in rat thoracic aorta.
PMID 24704610
Abstract
We previously showed that endothelin A (ETA) receptor antagonist BQ-123 partially inhibited cyclopiazonic acid (CPA)-enhanced endothelin-1 (ET-1)-induced contractions suggesting enhancement of ETA receptor internalization in caveolar structures by sarco/endoplasmic reticulum Ca+2 ATPase (SERCA) blockade. Since serotonin (5-Hydroxytryptamine, 5-HT) receptors are reported to be localized on caveolar membranes, we investigated whether SERCA inhibition affects 5-HT-induced responses and 5-HT receptor antagonism. For this purpose, vascular responses were measured in thoracic aorta segments from male Wistar albino rats using isolated tissue experiments. Data showed that CPA inhibits 5-HT- and PE-induced contractions in intact vessels while potentiating those in endothelium-denuded. Furthermore, non-selective 5-HT receptor blocker methysergide partially inhibited CPA-induced 5-HT contractions. However, α1-adrenergic receptor antagonist prazosin totally inhibited CPA-potentiated PE contractions. We suggest that SERCA inhibition results in 5-HT receptor internalization similar to ETA receptors possibly through protein kinase C activation by increased subsarcolemmal Ca2+ levels, eventually preventing 5-HT receptor antagonism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7313
1-[2-(Trifluoromethyl)phenyl]imidazole
C10H7F3N2