EMAIL THIS PAGE TO A FRIEND
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society
Natural compounds boldine and menthol are antagonists of human 5-HT3 receptors: implications for treating gastrointestinal disorders.
PMID 24708203
Abstract
Impaired 5-HT3 receptor function is likely involved in the pathogenesis of functional gastrointestinal disorders (FGID) and 5-HT3 receptor antagonists are effective treatments for chemotherapy-induced nausea and vomiting (CINV) and irritable bowel syndrome (IBS). The monoterpene alcohol menthol and the aporphine alkaloid boldine combat symptoms of gastrointestinal diseases; both interact with other members of the Cys-loop ligand-gated ion channel family and may therefore also act on 5-HT3 receptors. The impact of boldine and menthol on human recombinant homomeric 5-HT3 A- and heteromeric 5-HT3 AB receptors in HEK293 cells was determined by radioligand binding, a luminescence-based Ca(2+) assay, and a membrane potential assay. 5-HT3 protein and mRNA expression was assessed in human colon tissue. Boldine and menthol inhibited the 5-HT-induced activation of 5-HT3 receptors in the low and middle micromolar range, respectively. Boldine was a competitive antagonist of both receptors being 6.5- to 10-fold more potent at 5-HT3 A- vs 5-HT3 AB receptors. Menthol non-competitively and stereoselectively inhibited both receptors: In contrast to (+)-menthol, (-)-menthol was significantly more potent toward 5-HT3 A- vs 5-HT3 AB receptors. We show co-expression of 5-HT3A and 5-HT3B subunits in the human gut epithelium, the lamina propria, the myenteric plexus, and the muscular cell layer. The demonstrated 5-HT3 inhibitory effects may be relevant for boldine's and menthol's alleviating properties on FGID and may encourage clinical studies with the compounds or the plant extracts for CINV and IBS treatment. The found receptor-discriminative properties make boldine and (-)-menthol to potentially useful tools for analyzing structural differences between these receptor subtypes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
193984
1-Methylindole, ≥97%
C9H9N
H7752
Serotonin creatinine sulfate monohydrate, powder
C14H19N5O2 · H2O4S · H2O
H4511
Serotonin hydrogen maleate, powder
C10H12N2O · C4H4O4